Microbiological Analysis Filters
EMD Millipore™ EZ-Pak™ Membranes Available on GSA/VA Contract for Federal Government customers only.
Used for general microbiological analysis. Millipore™ EZ-Pak™ Membranes are sterile, ready-to-use membranes that eliminate contamination risk and save time.
EMD Millipore™ Microbiological Analysis Membrane Filters Available on GSA/VA Contract for Federal Government customers only.
Optimized for MF method microbiological analysis of water or other liquids
Fisherbrand™ Water-Testing Membrane Filters Available on GSA/VA Contract for Federal Government customers only.
Mixed Ester Cellulose membrane filters
Whatman™ Mixed Cellulose Ester Membranes - WME Range Available on GSA/VA Contract for Federal Government customers only.
Composed of cellulose acetate and cellulose nitrate membranes. Whatman™ Mixed Cellulose Ester Membranes - WME Range are characterized by a smoother and more uniform surface than pure nitrocellulose filters.
GVS Presterilized 47mm Membranes in 1+ PACs
Consistent porosity plus high strength
Whatman™ Mixed Cellulose Ester Membranes Available on GSA/VA Contract for Federal Government customers only.
Facilitates particle detection and minimizes eye fatigue with surface marks and color contrast. Whatman™ Mixed Cellulose Ester Membranes are available plain or gridded for automatic or manual counting procedures.
EMD Millipore™ Hydrophobic 3mm Edge Membranes Available on GSA/VA Contract for Federal Government customers only.
For microbiological and sterility testing of antibiotics and bacteriostatic drugs, as well as for filling machine operations
Whatman™ Mixed Cellulose Ester Membranes: ME24 Available on GSA/VA Contract for Federal Government customers only.
Recommended for the majority of routine applications
EMD Millipore™ Hydrophobic 6mm Edge Membranes Available on GSA/VA Contract for Federal Government customers only.
For microbiological and sterility testing of antibiotics and bacteriostatic drugs, as well as for filling machine operations
EMD Millipore™ EZ-Pak™ Membrane Dispenser Curve (AC only)
Used for general microbiological analysis. EMD Millipore EZ-Pak™ Membrane Dispenser Curve (AC only) is without battery. It is fast, robust, and has no-touch dispensing operation.
Whatman™ Cellulose Nitrate Membrane - Grade NC45 Available on GSA/VA Contract for Federal Government customers only.
Featutring very narrow pore size distribution and low levels of extractable
Whatman™ Non-sterile ME 25 Mixed Cellulose Ester Membranes Available on GSA/VA Contract for Federal Government customers only.
Offers a larger adsorption of product with a high degree of internal surface area. Whatman™ Non-sterile ME25 Mixed Cellulose Membranes are available plain for automatic colony counting applications, as well as gridded to assist in manual counting procedures.
Whatman™ MicroPlus Membranes Available on GSA/VA Contract for Federal Government customers only.
Features very narrow pore size distribution, low levels of extractables and increased temperature stability. Whatman™ MicroPlus Membranes are suitable for sample preparation, microbiological studies and filtration of aqueous solutions.
Whatman™ Grade GF/B Glass Microfiber Filters Available on GSA/VA Contract for Federal Government customers only.
For liquid scintillation counting and liquid clarification for suspended solids analysis.
EMD Millipore™ Milliwrap™ and Milliseal™ Adhesive Membranes Available on GSA/VA Contract for Federal Government customers only.
Convenient replacements for metal caps, parafilm, cotton plugs and lids on laboratory containers
EMD Millipore™ EZ-Pak™ Dispenser
Sterile, ready-to-use membranes eliminate contamination risk, save time
Sartorius™ Microsart™ e.motion™ Membrane Dispenser Available on GSA/VA Contract for Federal Government customers only.
Versatile and flexible fully automated membrane filter dispenser for individually, sterile-packaged cellulose nitrate membrane filter discs
  spinner