Stains and Dyes for Microbiology
Thermo Scientific™ Remel™ Gram Stain Kit, 4 x 250 mL bottles Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Gram Stain Kit is for qualitative procedures to differentiate gram-positive from gram-negative microorganisms.
Thermo Scientific™ Remel™ Gram Decolorizer Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Gram Decolorizer is a decolorizing reagent used in Gram stain procedures.
eBioscience™ Fixable Viability Dye eFluor™ 506 Non-distribution product as customer accommodation.
Can be used to irreversibly label dead cells prior to cryopreservation, fixation and/or permeabilization procedures. Unlike 7-AAD and PI, cells labeled with Fixable Viability Dyes can be washed, fixed, permabilized and stained for intracellular antigens without loss of staining intensity.
BD BBL™ Gram Stain Kits Available on GSA/VA Contract for Federal Government customers only.
Used to stain microorganisms from cultures or specimens by the differential Gram method
BD Oxidase Reagent Dropper Available on GSA/VA Contract for Federal Government customers only.
Identify microorganisms without mixing or measuring reagents
BD DrySlide™ Reagent Slides Available on GSA/VA Contract for Federal Government customers only.
A convenient method for bacterial biochemical analysis. Self-contained, ready-to-use test slides consist of coated, 35mm film or impregnated paper, which operates as a carrier matrix for microbiological testing reagents.
BD Gram Decolorizer Available on GSA/VA Contract for Federal Government customers only.
Can be used as a replacement reagent for the BD™ Gram Stain Kits. BD™ Gram Decolorizer is used in a 4-step Gram staining process, before counterstaining. Available in 250ml solution bottles for direct use or gallon containers for bulk storage.
BD DMACA Indole Reagent Available on GSA/VA Contract for Federal Government customers only.
Identify microorganisms without mixing or measuring reagents
Fisher HealthCare™ PROTOCOL™ Gram Stain Sets
Reusable tray provides secure bottle storage
Thermo Scientific™ Remel™ Gram Safranin Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Gram Safranin is a counterstain used in Gram stain procedures.
BD Gram Crystal Violet Available on GSA/VA Contract for Federal Government customers only.
Can be used as a replacement reagent for the BD™ Gram Stain Kits. BD™ Gram Crystal Violet is used as the primary stain in a 4-step Gram staining process. Available in 250ml solution bottles for direct use or gallon containers for bulk storage.
BD Difco™ BBL™ Stains, Dyes, and Indicators
For Gram stain and acid-fast staining procedures
Thermo Scientific™ Remel™ Gram Iodine Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Gram Iodine is a mordant used in Gram stain procedures.
BD Catalase Reagent Dropper Available on GSA/VA Contract for Federal Government customers only.
Identify microorganisms without mixing or measuring reagents
BD Indole Reagent Dropper Available on GSA/VA Contract for Federal Government customers only.
Identify microorganisms without mixing or measuring reagents
Fisherbrand™ PROTOCOL™ Individual Gram Stains Available on GSA/VA Contract for Federal Government customers only.
Manufactured with traditional formulations used by most laboratories
BD Fixable Viability Stain 780
Useful for discrimination of viable from non-viable mammalian cells in multicolor flow cytometric applications. BD Horizon™ Fixable Viability Stain 780 reacts with and covalently binds to cell-surface and intracellular amines.
BD Voges Proskauer B Reagent Dropper
Identify microorganisms without mixing or measuring reagents
Thermo Scientific™ Remel™ Gram Crystal Violet Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Gram Crystal Violet is a primary stain in Gram stain procedures.
BD Voges Proskauer A Reagent Dropper
Identify microorganisms without mixing or measuring reagents
BD Nitrate B Reagent Dropper
Identify microorganisms without mixing or measuring reagents
BD Nitrate A Reagent Dropper
Identify microorganisms without mixing or measuring reagents
BD BBL™/Difco™ TB Carbolfuchsin KF Available on GSA/VA Contract for Federal Government customers only.
For staining mycobacteria by the Kinyoun (cold) acid-fast procedure
BD 10% Potassium Hydroxide Reagent Dropper Available on GSA/VA Contract for Federal Government customers only.
Identify microorganisms without mixing or measuring reagents
Thermo Scientific™ Remel™ Trichrome Quick Stain Kit Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Trichrome Quick Stain Kit is for use with mercury-free transports.
BD BBL™/Difco™ TB Stain Kit ZN
For staining mycobacteria by the Ziehl-Neelsen (hot) acid-fast procedure
Eng Scientific Cryptosporidium Stain Kit
Specimens may be prepared from fresh material or preserved in 10% buffered formalin
Thermo Scientific™ Remel™ TB Kinyoun Carbolfuchsin Stain Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel TB Kinyoun Carbolfuchsin is a primary stain for AFB and Cryptosporidium modified acid-fast staining procedures.
Thermo Scientific™ Remel™ TB Auramine-Rhodamine T Stain Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel TB Auramine-Rhodamine T is a fluorescent stain for microscopic detection of AFB.
  spinner