Microbiology Stains Dyes and Indicators
Thermo Scientific™ Remel™ Gram Stain Kit, 4 x 250 mL bottles Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Gram Stain Kit is for qualitative procedures to differentiate gram-positive from gram-negative microorganisms.
Thermo Scientific™ Remel™ Gram Iodine Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Gram Iodine is a mordant used in Gram stain procedures.
BD Oxidase Reagent Dropper Available on GSA/VA Contract for Federal Government customers only.
Identify microorganisms without mixing or measuring reagents
BD DrySlide™ Reagent Slides Available on GSA/VA Contract for Federal Government customers only.
A convenient method for bacterial biochemical analysis. Self-contained, ready-to-use test slides consist of coated, 35mm film or impregnated paper, which operates as a carrier matrix for microbiological testing reagents.
BD BBL™ Gram Stain Kits Available on GSA/VA Contract for Federal Government customers only.
Used to stain microorganisms from cultures or specimens by the differential Gram method
eBioscience™ Fixable Viability Dye eFluor™ 506 Non-distribution product as customer accommodation.
Can be used to irreversibly label dead cells prior to cryopreservation, fixation and/or permeabilization procedures. Unlike 7-AAD and PI, cells labeled with Fixable Viability Dyes can be washed, fixed, permabilized and stained for intracellular antigens without loss of staining intensity.
BD Catalase Reagent Dropper Available on GSA/VA Contract for Federal Government customers only.
Identify microorganisms without mixing or measuring reagents
BD Gram Decolorizer Available on GSA/VA Contract for Federal Government customers only.
Can be used as a replacement reagent for the BD™ Gram Stain Kits. BD™ Gram Decolorizer is used in a 4-step Gram staining process, before counterstaining. Available in 250ml solution bottles for direct use or gallon containers for bulk storage.
BD Voges Proskauer A Reagent Dropper
Identify microorganisms without mixing or measuring reagents
BD Difco™ BBL™ Stains, Dyes, and Indicators
For Gram stain and acid-fast staining procedures
BD Nitrate A Reagent Dropper
Identify microorganisms without mixing or measuring reagents
Thermo Scientific™ Remel™ Gram Decolorizer Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Gram Decolorizer is a decolorizing reagent used in Gram stain procedures.
BD Nitrate B Reagent Dropper
Identify microorganisms without mixing or measuring reagents
Fisher HealthCare™ PROTOCOL™ Gram Stain Sets
Reusable tray provides secure bottle storage
BD Voges Proskauer B Reagent Dropper
Identify microorganisms without mixing or measuring reagents
Fisher HealthCare™ PROTOCOL™ Individual Gram Stains Available on GSA/VA Contract for Federal Government customers only.
Manufactured with traditional formulations used by most laboratories
BD Indole Reagent Dropper Available on GSA/VA Contract for Federal Government customers only.
Identify microorganisms without mixing or measuring reagents
BD Fixable Viability Stain 780
Useful for discrimination of viable from non-viable mammalian cells in multicolor flow cytometric applications. BD Horizon™ Fixable Viability Stain 780 reacts with and covalently binds to cell-surface and intracellular amines.
BD Gram Crystal Violet Available on GSA/VA Contract for Federal Government customers only.
Can be used as a replacement reagent for the BD™ Gram Stain Kits. BD™ Gram Crystal Violet is used as the primary stain in a 4-step Gram staining process. Available in 250ml solution bottles for direct use or gallon containers for bulk storage.
Thermo Scientific™ Remel™ Gram Safranin Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Gram Safranin is a counterstain used in Gram stain procedures.
BD Horizon™ Fixable Viability Stain 510
Useful for discrimination of viable from non-viable mammalian cells in multicolor flow cytometric applications
BD DMACA Indole Reagent Available on GSA/VA Contract for Federal Government customers only.
Identify microorganisms without mixing or measuring reagents
Thermo Scientific™ Remel™ Gram Iodine PVP Available on GSA/VA Contract for Federal Government customers only.
Differs from traditional Gram stain formulation by substituting PVP iodine, an organic iodine complex, which is more stable than iodine.
Thermo Scientific™ Remel™ Gram Crystal Violet Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Gram Crystal Violet is a primary stain in Gram stain procedures.
BD 10% Potassium Hydroxide Reagent Dropper Available on GSA/VA Contract for Federal Government customers only.
Identify microorganisms without mixing or measuring reagents
Corning™ Trypan Blue Available on GSA/VA Contract for Federal Government customers only.
Can be used for variety cell counting methods requiring information on cell count and viability.
BD Fixable Viability Stain 450
Useful to discriminate viable from non-viable mammalian cells in multicolor flow cytometric application
Thermo Scientific™ Remel™ BactiDrop Calcofluor White Available on GSA/VA Contract for Federal Government customers only.
A fluorescent stain for fungi, Microsporidium, Acanthamoeba, and Pneumocystis.
BD BBL™/Difco™ TB Stain Kit K Available on GSA/VA Contract for Federal Government customers only.
For staining mycobacteria by the Kinyoun (cold) acid-fast procedure
  spinner