MIC Test Strips
Eisco™ N-Propylthiouracil Test Paper, 100 Strips Products for Science Education
N-Propylthiouracil test paper tests for the genetically controlled ability to taste N-Propylthiouraci (PROP) . If the strip tastes bitter to an individual they have the homozygous dominant or heterozygous condition. Supplied in a vial of 100 strips
  spinner