Nucleic Acid Purification
Thermo Scientific™ KingFisher™ Flex™ Systems Consumables
Optimize performance using consumables specifically designed for use with the KingFisher™ Flex™ System and ideal for magnetic particle processing.
Thermo Scientific™ GeneJET™ Plasmid Miniprep Kit Available on GSA/VA Contract for Federal Government customers only.
Perform isolation of high quality plasmid DNA from recombinant E. coli cultures with this simple, rapid, and cost-effective system.
Invitrogen™ TRIzol™ Reagent Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Isolate RNA, DNA, and protein from the same sample
Invitrogen™ RNAlater™ Stabilization Solution Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Immediate RNase inactivation and RNA stabilization within tissues or cells
Invitrogen™ UltraPure™ UltraPure Agarose Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Easy-pour spout reduces spills and contamination
Invitrogen™ RNaseOUT™ Recombinant Ribonuclease Inhibitor Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Protects mRNA and improves total cDNA yields
Applied Biosystems™ MagMAX™ DNA Multi-Sample Ultra Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Optimized to isolate genomic DNA (gDNA) from a variety of samples, such as whole blood, buccal cells, saliva, urine, blood cards, mouth rinse, and tissue
Invitrogen™ SUPERase·In™ RNase Inhibitor (20U/μL) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Binds and inactivates a wide variety of RNases
Invitrogen™ BenchPro™ 2100 Plasmid Purification Card and Reagent Tray Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used with the BenchPro™ 2100 Plasmid Processing Station for large-scale purification of plasmid DNA from bacterial cells
Invitrogen™ MagMAX™-96 Total RNA Isolation Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Magnetic bead-based technology to purify total RNA
Applied Biosystems™ DNA Extract All Reagents Kit Non-distribution product as customer accommodation.
Provides PCR-ready DNA from a wide variety of sample types, ranging from blood to buccal swabs to plant tissues, in 5 minutes
Applied Biosystems™ Tempus™ Blood RNA Tube Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Stabilize RNA and lyse whole blood cells in one step
Invitrogen™ PureLink™ RNA Mini Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Provides a simple, reliable, and rapid column-based method for isolating high-quality total RNA from a wide variety of sources without the need for hazardous reagents such as phenol
Thermo Scientific™ KingFisher™ Duo Consumables
Carry out magnetic particle processing with Thermo Scientific™ KingFisher™ Duo and KingFisher Duo Prime Consumables, specially designed combs and microplates optimized for performance.
MP Biomedicals™ FastPrep-24™ 5G Instrument
Delivering the most DNA, RNA and Proteins from the most resistant samples in less than 40 seconds.  MP Biomedicals™ FastPrep-24™ 5G Instrument  is a high-speed benchtop homogenizer.
Invitrogen™ PureLink™ Genomic DNA Mini Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
No detectable contamination from carryover of protein or guanidine
MP Biomedicals™ Lysing Matrix D Available on GSA/VA Contract for Federal Government customers only.
FastPrep™ Lysing Matrix D are used primarily with FastPrep™ homogenizers or any other bead beater, for lysis of softer tissues like brain, liver, kidney, lung and spleen. Also used to lyse plant tissue such as leaves, roots, fruits as well as cultured cells and whole insects.
Applied Biosystems™ RNase Inhibitor Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
No DNase or endonuclease activity
Thermo Scientific™ GeneJET™ Gel Extraction Kit Available on GSA/VA Contract for Federal Government customers only.
Perform silica membrane-based DNA fragment purification from standard or low-melting point agarose gels run in either TAE or TBE buffer.
Invitrogen™ TRIzol™ LS Reagent Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Recovery RNA, DNA, and protein from a single liquid sample
Thermo Scientific™ KingFisher™ Systems Consumables
Optimize performance using Thermo Scientific™ KingFisher™Systems Consumables, ideal for magnetic particle processing.
Applied Biosystems™ ExoSAP-IT™ PCR Product Clean-up Reagent Non-distribution product as customer accommodation.
ExoSAP-IT™ PCR Product Clean-up Reagent is used for enzymatic clean-up of amplified PCR product. It hydrolyses excess primers and nucleotides in a single step.
Invitrogen™ TRIzol™ Plus RNA Purification Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Isolate total RNA from animal and plant cells, tissue, bacteria, and yeast
Thermo Scientific™ Consumables for KingFisher™ mL Systems
Optimize system performance with Consumables for Thermo Scientific™ KingFisher™ mL Systems.
Invitrogen™ TE Buffer Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Component in PureLink 96 Plasmid Purification System
Invitrogen™ mirVana™ miRNA Isolation Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Available with and without phenol
MP Biomedicals™ Lysing Matrix B, 2mL tubes Available on GSA/VA Contract for Federal Government customers only.
Contains 0.1mm silica spheres. Used primarily for lysis of gram positive and gram negative bacteria, and will also disrupt fungal tissue and spores. Lysing Matrix B can also be used as an additive for disruption by sonication.
  spinner