RNA Isolation and Purification Products
MP Biomedicals™ Lysing Matrix D Available on GSA/VA Contract for Federal Government customers only.
FastPrep™ Lysing Matrix D are used primarily with FastPrep™ homogenizers or any other bead beater, for lysis of softer tissues like brain, liver, kidney, lung and spleen. Also used to lyse plant tissue such as leaves, roots, fruits as well as cultured cells and whole insects.
MP Biomedicals™ Lysing Matrix B, 2mL tubes Available on GSA/VA Contract for Federal Government customers only.
Contains 0.1mm silica spheres. Used primarily for lysis of gram positive and gram negative bacteria, and will also disrupt fungal tissue and spores. Lysing Matrix B can also be used as an additive for disruption by sonication.
MP Biomedicals™ Lysing Matrix A, 2mL tubes Available on GSA/VA Contract for Federal Government customers only.
For all sample types except soil for the subsequent isolation of genomic DNA. MP Biomedicals Lysing Matrix A impact-resistant tubes contains garnet matrix and one 1/4 in. ceramic sphere.
Promega Maxwell™ RSC simplyRNA Tissue Kit
Automated RNA purification from tissues using a low elution volume using the Maxwell™ RSC Instrument. The Maxwell™ RSC simplyRNA Tissue Kit provides the necessary reagents for processing samples and uses prefilled cartridges for purification, maximizing simplicity and convenience.
MP Biomedicals™ Lysing Matrix E, 2mL tubes Available on GSA/VA Contract for Federal Government customers only.
Lysing Matrix E is used primarily for environmental samples such as soil, sludge, wastwater and feces. Lysing Matrix E can also be effective in lysing mixed samples, such as microbe infected tissue, and can be used in RNA extraction from tumors and other difficult tissues.
Promega Maxwell™ RSC simplyRNA Blood Kit
Automated purification of RNA from whole blood using a low elution volume with the Maxwell™ RSC Instrument. The kit provides the necessary reagents for processing the samples and uses prefilled cartridges for purification, maximizing simplicity and convenience.
MP Biomedicals™ Lysing Matrix S, 2mL tubes Available on GSA/VA Contract for Federal Government customers only.
Use to break hard or brittle samples with minimal amounts of mechanical shearing. Specialty applications include RNA and protein extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.
Promega Maxwell™ RSC RNA FFPE Kit
Automated RNA extraction from FFPE tissue sections using the Maxwell™ RSC Instrument. The kit's predispensed reagent cartridges are designed to work with the instrument's preprogrammed purification procedures, maximizing simplicity and convenience.
Promega Maxwell™ RSC simplyRNA Cells Kit
Automated RNA ourification from cells using a low elution volume with the Maxwell™ RSC Instrument. The kit provides the necessary reagents for processing the samples and uses prefilled cartridges for purification, maximizing simplicity and convenience.
MP Biomedicals™ FastRNA™ SPIN RNA Isolation Kit Available on GSA/VA Contract for Federal Government customers only.
Isolate total RNA from bacterial cell cultures and other microbial sources in approximately 15 minutes. Does not use phenol or reducing agents.
MP Biomedicals™ Lysing Matrix A, 15mL tubes Available on GSA/VA Contract for Federal Government customers only.
For all sample types except soil for the subsequent isolation of genomic DNA. Each impact-resistant 15mL tube contains garnet matrix and ten 1/4 in. ceramic sphere. Lysing Matrix A is chemically inert and will not bind nucleic acids.
Promega Maxwell™ 16 LEV Plant RNA Kit
Automated RNA extraction from 1-16 plant tissue samples in 60 minutes using the Maxwell™ 16 Research Instrument. The Maxwell™ 16 LEV Plant RNA Kit supplies predispensed reagent cartridges that work with the instrument's preprogrammed settings.
Promega™ QuantiFluor™ RNA System
Easily detects and quantifies small amounts of RNA in solution using a single-tube or plate reading fluorometer, supplied with QuantiFluor RNA Dye, Dilution Buffer and RNA standard
Promega Maxwell™ 16 miRNA Tissue Kit
Automated extraction of high-quality total RNA with enhanced miRNA enrichment using the Maxwell™ 16 MDx Instrument. The kit contains all the necessary reagents for miRNA extraction in a convenient prefilled cartridge format.
MP Biomedicals™ Lysing Matrix Z Available on GSA/VA Contract for Federal Government customers only.
Used primarily for lysis of animal and plant tissue, tougher stems, roots and seeds, and whole insectsDNase and RNase free.
Promega Maxwell™ CSC DNA FFPE Kit
Automated DNA purification from FFPE samples for In Vitro diagnostic assays using the Maxwell™ CSC Instrument.
MP Biomedicals™ Lysing Matrix J Available on GSA/VA Contract for Federal Government customers only.
Used for nucleic acid isolation and purification
MP Biomedicals™ TallPrep™ Tubes
For use in Nucleic Acid Isolation and Purification
MP Biomedicals™ Lysing Matrix C, 2mL tubes Available on GSA/VA Contract for Federal Government customers only.
Contains 1mm silica spheres. Lysing Matrix C is used primarily for lysis of yeast and fungi and can also be used for molds, spores and filamentous algae.
MP Biomedicals™ Lysing Matrix M Available on GSA/VA Contract for Federal Government customers only.
Tubes for optimal lysis used with FastPrep™ instruments
MP Biomedicals™ Lysing Matrix B, 96-well racks (1.2mL) Available on GSA/VA Contract for Federal Government customers only.
Contains 0.1mm silica spheres. Used primarily for lysis of gram positive and gram negative bacteria, and will also disrupt fungal tissue and spores. Lysing Matrix B can also be used as an additive for disruption by sonication.
MP Biomedicals™ Lysing Matrix S, 96-Well, 1.2mL
MP Biomedicals™ Lysing Matrix S, 96-Well, 1.2mL extractions of nucleic acids, proteins, metabolites and other small molecules from tough, hard or brittle cell membranes including organisms with a tough or dense exterior matrix.
MP Biomedicals™ Lysing Matrix G Available on GSA/VA Contract for Federal Government customers only.
Tubes for optimal lysis used with FastPrep™ instruments
MP Biomedicals™ FastRNA™ Pro Soil-Direct Kit Available on GSA/VA Contract for Federal Government customers only.
Used to isolate total cellular RNA from microorganisms and other specimens found in soil samples. Designed for use with FastPrep™ Instrument. Two levels of RNA purification, for different soil samples and downstream applications.
MilliporeSigma™ Novagen™ RNase A Solution
Suitable for use in selective removal of RNA.
Promega Maxwell™ 16 LEV simplyRNA Blood Kit
Simple, automated RNA extraction
Promega ReliaPrep RNA Cell Miniprep System, 50 preps
The ReliaPrep RNA Miniprep Systems provide a fast and simple technique for preparation of intact total RNA from cultured cells or tissue in as little as 30 minutes from very small amounts of input material.
Ambion™ Pre-miR™ miRNA Precursor Non-distribution product as customer accommodation.
Pre-miR™ miRNA Precursor
  spinner