DNA Synthesis Reagents and Columns
Thermo Scientific™ RevertAid™ Premium Double Stranded cDNA Synthesis Kit
The Thermo Scientific Maxima H Minus Double-Stranded cDNA Synthesis Kit is a complete system for efficient synthesis of double-stranded cDNA from total RNA or mRNA.
Applied Biosystems™ Triethylamine Acetate (TEAA) 2.0 M Non-distribution product as customer accommodation.
This 2.0 M Triethylamine Acetate solution is used with OPC™ cartridges (Cat. No. 400771), for oligonucleotide purification.
Applied Biosystems™ Trap-Pak™ Bags (Medium) Non-distribution product as customer accommodation.
Trap-Pak™ bags are tightly sealed membrane packets containing molecular sieves to reduce and maintain a low water content in solutions that must remain anhydrous.
Invitrogen™ GeneArt™ High-Order Genetic Assembly System Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
A highly efficient kit for the simultaneous and seamless assembly of up to 10 DNA fragments, totaling up to 110 Kbp in length, into any vector.
Applied Biosystems™ Trap-Pak™ Bags (Mini) Non-distribution product as customer accommodation.
Trap-Pak™ bags are tightly sealed membrane packets containing molecular sieves to reduce and maintain a low water content in solutions that must remain anhydrous.
Invitrogen™ GeneArt™ Seamless Cloning and Assembly Enzyme Mix Non-distribution product as customer accommodation.
Enables the simultaneous and directional cloning of 1 to 4 PCR fragments, consisting of any sequence, into any linearized vector, in a single 30-minute room temperature reaction
Invitrogen™ GeneArt™ Seamless PLUS Cloning and Assembly Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
The complete kit for simultaneous and directional cloning of 1 to 4 PCR fragments, consisting of any sequence, into any linearized vector, in a single 30-minute or less, room temperature reaction
Invitrogen™ GeneArt™ pYES1L Vector with Sapphire™ Technology Non-distribution product as customer accommodation.
A ready-to-use, 9.3 Kb linearized BAC⁄YAC shuttle vector, used for the assembly of DNA fragments with the GeneArt™ High-Order Genetic Assembly System
Invitrogen™ GeneArt™ Seamless Cloning and Assembly Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Enables the simultaneous and directional cloning of 1 to 4 PCR fragments, consisting of any sequence, into any linearized vector, in a single 30-minute room temperature reaction
Invitrogen™ GeneArt™ Type IIs Assembly Kit Non-distribution product as customer accommodation.
Provides seamless cloning and assembly of up to 8 DNA fragments by simultaneous cleavage and ligation in a single reaction
Invitrogen™ GeneArt™ High-Order Vector Conversion Cassette Non-distribution product as customer accommodation.
A ready-to-use, 3.8kb linear BAC⁄YAC cassette, containing yeast replication and selection elements that can make any E.coli vector compatible with the GeneArt™ High-Order Genetic Assembly System
Thermo Scientific™ TransformAid Bacterial Transformation Kit Promotion
Kit for quick and convenient preparation of competent E.coli cells for guaranted transformation efficiencies of more than 107 transformants per µg of plasmid DNA.
Thermo Scientific™ Fast DNA End Repair Kit Promotion
Perform blunting and phosphorylation of DNA ends in just 5 minutes for subsequent use in blunt-end ligation with this kit.
Transgenomic CE Phosphoramidites
High-quality, low-cost DNA synthesis reagents compatible with most automated synthesizers
  spinner