Unlabeled Oligonucleotides and Primers
Applied Biosystems™ Random Hexamers (50mM) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Serve as primers for DNA synthesis by a DNA polymerase or reverse transcriptase
Thermo Scientific™ Primers for cDNA Synthesis Available on GSA/VA Contract for Federal Government customers only.
Optimze cDNA synthesis with these random hexamer primers, oligo(dT)18 primers and anchored oligo dT primers.
Invitrogen™ Random Primers Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Truly random primers suitable for DNA synthesis using Klenow fragments with DNA templates or for cDNA synthesis using reverse transcriptase with mRNA templates
Invitrogen™ Oligo(dT) 20 Primer Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used for first-strand cDNA synthesis with reverse transcriptase at temperatures of ≥50°C
Invitrogen™ Oligo(dT) 12-18 Primer Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Primer is suitable for use in first-strand cDNA synthesis with reverse transcriptase
Invitrogen™ Oligo (dT) Primer (50mM) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Provided at a stock concentration of 50μM
Invitrogen™ Random Decamers (50μM) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Provided at a stock concentration of 50μM, and functionally tested using the RETROscript™ Kit
Thermo Scientific™ Oligo(dT)18 Primers Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Oligo(dT)18 Primer is a synthetic single-stranded 18-mer oligonucleotide with 5'- and 3'-hydroxyl ends.
Invitrogen™ Bio-16-UTP (10mM) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Ideal for use as substrates as part of in vitro transcription reactions
Promega Random and Oligo (dT) Primers
For first-strand cDNA synthesis and cloning
Invitrogen™ Bio-11-UTP (75mM) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Ideal for use as substrates as part of in vitro transcription reactions
Applied Biosystems™ Oligo d(T) 16 (50mM) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used for priming and reverse transcription of polyadenylated (poly A+) mRNA
Invitrogen™ 5-(3-Aminoallyl)-dUTP (50mM) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
When incorporated into DNA, 5-(3-aminoallyl)-dUTP provides a reactive group for addition of other chemical groups
Invitrogen™ Bio-11-UTP (10mM) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Ideal for use as substrates as part of in vitro transcription reactions
Thermo Scientific™ Exo-Resistant Random Primer Available on GSA/VA Contract for Federal Government customers only.
Perform highly efficient random priming of DNA synthesis reactions with this mixture of single-stranded random oligonucleotides.
Invitrogen™ Anchored Oligo(dT) 20 Primer Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Primer mixture consisting of a string of 20 deoxythymidylic acid residues followed by dV (either dG, dA, or dC) and then by dN (dA, dT, dG, or dC)
Invitrogen™ RNA Century™-Plus Marker Templates Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Contain mixtures of linearized plasmids ready for use as templates during in vitro transcription reactions for synthesis of labeled RNA molecular size standards
Invitrogen™ Lambda Hind III dsDNA Markers Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Ambion Lambda DNA is digested to completion with Hind III
Thermo Scientific™ M13/pUC sequencing primer (-20), 17-mer Available on GSA/VA Contract for Federal Government customers only.
Accurately sequence DNA with M13 pUC sequencing primers, single-stranded oligonucleotides with 5'-hydroxyl and 3'-hydroxyl ends.
Invitrogen™ RNA Century™ Marker Templates Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Contain mixtures of linearized plasmids ready for use as templates as part of in vitro transcription reactions for synthesis of labeled RNA molecular size standards
Thermo Scientific™ Transcription Promoter Sequencing Primers Available on GSA/VA Contract for Federal Government customers only.
Perform sequencing of DNA fragments located downstream from the corresponding RNA polymerase promoter sequences in cloning vectors, such as pTZ19R, pTZ57R and pBluescript II.
Thermo Scientific™ pJET1.2 Sequencing Primers Available on GSA/VA Contract for Federal Government customers only.
Primers for sequencing of DNA fragments inserted into Eco32I site within the eco47IR gene of pJET1.2 and for colony screening by PCR.
Thermo Scientific™ Transcription Promoter Sequencing Primers Available on GSA/VA Contract for Federal Government customers only.
Perform sequencing of DNA fragments located downstream from the corresponding RNA polymerase promoter sequences in cloning vectors, such as pTZ19R, pTZ57R and pBluescript II.
Thermo Scientific™ M13/pUC reverse sequencing primer (-46), 24-mer Available on GSA/VA Contract for Federal Government customers only.
Sequence DNA fragments inserted into the MCS of various pUC19 based cloning vectors with M13 pUC sequencing primers, single-stranded oligonucleotides.
Thermo Scientific™ Transcription Promoter Sequencing Primers Available on GSA/VA Contract for Federal Government customers only.
Perform sequencing of DNA fragments located downstream from the corresponding RNA polymerase promoter sequences in cloning vectors, such as pTZ19R, pTZ57R and pBluescript II.
Thermo Scientific™ Transcription Promoter Sequencing Primers Available on GSA/VA Contract for Federal Government customers only.
Perform sequencing of DNA fragments located downstream from the corresponding RNA polymerase promoter sequences in cloning vectors, such as pTZ19R, pTZ57R and pBluescript II.
Thermo Scientific™ Transcription Promoter Sequencing Primers Available on GSA/VA Contract for Federal Government customers only.
Perform sequencing of DNA fragments located downstream from the corresponding RNA polymerase promoter sequences in cloning vectors, such as pTZ19R, pTZ57R and pBluescript II.
  spinner