DNA Extraction and Purification
Thermo Scientific™ GeneJET™ Plasmid Miniprep Kit Available on GSA/VA Contract for Federal Government customers only.
Perform isolation of high quality plasmid DNA from recombinant E. coli cultures with this simple, rapid, and cost-effective system.
Invitrogen™ UltraPure™ UltraPure Agarose Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Easy-pour spout reduces spills and contamination
Applied Biosystems™ MagMAX™ DNA Multi-Sample Ultra Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Optimized to isolate genomic DNA (gDNA) from a variety of samples, such as whole blood, buccal cells, saliva, urine, blood cards, mouth rinse, and tissue
Invitrogen™ PureLink™ Genomic DNA Mini Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
No detectable contamination from carryover of protein or guanidine
Applied Biosystems™ DNA Extract All Reagents Kit Non-distribution product as customer accommodation.
Provides PCR-ready DNA from a wide variety of sample types, ranging from blood to buccal swabs to plant tissues, in 5 minutes
Thermo Scientific™ GeneJET™ Gel Extraction Kit Available on GSA/VA Contract for Federal Government customers only.
Perform silica membrane-based DNA fragment purification from standard or low-melting point agarose gels run in either TAE or TBE buffer.
Invitrogen™ TE Buffer Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Component in PureLink 96 Plasmid Purification System
GE Healthcare illustra™ NAP™ Columns, NAP-5 Available on GSA/VA Contract for Federal Government customers only.
Use for rapid and efficient purification of DNA and oligonucleotides (>10-mer) in less than 15 minutes by gravity flow. GE Healthcare Life Sciences™ illustra™ NAP™ Columns are disposable columns prepacked with Sephadex™ G-25 DNA Grade
Invitrogen™ PureLink™ Quick Plasmid Miniprep Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Contaminants are removed with Wash Buffer
Invitrogen™ PureLink™ HiPure Plasmid Maxiprep Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
No organic solvents, ethidium bromide, or cesium chloride
Invitrogen™ UltraPure™ Salmon Sperm DNA Solution Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used directly in preparation of prehybridization and hybridization solutions
Promega™ Maxwell™ 16 Purification Kits
Used with Maxwell 16 Instruments to provide an easy method for efficient, automated purification of genomic DNA
MP Biomedicals™ 1/4 in. Ceramic Sphere Available on GSA/VA Contract for Federal Government customers only.
Use to break hard/brittle samples. Specialty applications for the 1/4 in. Ceramic Sphere include RNA extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.
Invitrogen™ UltraPure™ 1M Tris-HCI Buffer, pH 8.0 Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Pre-mixed, pH-adjusted, sterile-filtered solutions
Promega Wizard™ Plus SV Minipreps DNA Purification System
Produces high yield plasmid DNA of consistent quality from bacterial cultures
Promega™ Wizard™ SV Gel and PCR Cleanup System
Extract DNA fragments from standard or low-melt agarose gels or purify PCR products directly from an amplification reaction. Purified DNA can be used for automated fluorescent DNA sequencing, cloning, labeling, restriction enzyme digestion, or in vitro transcription/translation.
Invitrogen™ PureLink™ HiPure Plasmid Midiprep Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
No organic solvents, ethidium bromide, or cesium chloride
GE Healthcare illustra™ NAP™ Columns, NAP-25 Available on GSA/VA Contract for Federal Government customers only.
Use for rapid and efficient purification of DNA and oligonucleotides (>10-mer) in less than 15 minutes by gravity flow. GE Healthcare Life Sciences™ illustra™ NAP™ Columns are disposable columns prepacked with Sephadex™ G-25 DNA Grade
Thermo Scientific™ GeneJET™ PCR Purification Kit Available on GSA/VA Contract for Federal Government customers only.
Perform rapid and efficient purification of DNA from PCR and other enzymatic reaction mixtures with this kit.
GE Healthcare illustra™ MicroSpin™ G-25 Columns Available on GSA/VA Contract for Federal Government customers only.
Use for the rapid purification of DNA >10 base pairs in length in a wide range of applications. GE Healthcare Life Sciences™ llustra™ MicroSpin™ G-25 Columns are ideal for desalting, buffer exchange, and removal of unincorporated nucleotides from end-labeled oligonucleotides. 
Invitrogen™ UltraPure™ 1 M Tris-HCI Buffer, pH 7.5 Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Pre-mixed, pH-adjusted, sterile-filtered solution
Thermo Scientific™ GeneJET™ Plasmid Maxiprep Kit Available on GSA/VA Contract for Federal Government customers only.
Perform large-scale isolation of high-quality plasmid DNA from recombinant E. coli cultures with this kit.
GE Healthcare illustra™ NAP™ Columns, NAP-10 Available on GSA/VA Contract for Federal Government customers only.
Use for rapid and efficient purification of DNA and oligonucleotides (>10-mer) in less than 15 minutes by gravity flow. GE Healthcare Life Sciences™ illustra™ NAP™ Columns are disposable columns prepacked with Sephadex™ G-25 DNA Grade
Axygen™ AxyPrep Mag™ PCR Clean-up Kits Available on GSA/VA Contract for Federal Government customers only.
Post PCR clean-up simplified
MP Biomedicals™ FastDNA™ SPIN Kit for Soil Available on GSA/VA Contract for Federal Government customers only.
Isolates DNA from cells present in soil or other environmental samples.   MP Biomedicals™ FastDNA™ SPIN Kit for Soil extracts DNA easily in a 2mL tube.
  spinner