DNA Polymerases
Thermo Scientific™ DreamTaq Green PCR Master Mix (2X) Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific DreamTaq Green PCR Master Mix (2X) is a ready-to-use solution containing DreamTaq DNA Polymerase, optimized DreamTaq Green buffer, MgCl2, and dNTPs.
Thermo Scientific™ Maxima™ H Minus Reverse Transcriptase Available on GSA/VA Contract for Federal Government customers only.
A highly thermostable, high processivity reverse transcriptase lacking RNase H activity.
Thermo Scientific™ DreamTaq Green PCR Master Mix (2X) Available on GSA/VA Contract for Federal Government customers only.
Save time in PCR applications with Master Mix and get higher sensitivity, longer PCR products and higher yields compared to conventional Taq DNA polymerase.
Invitrogen™ SuperScript™ IV Reverse Transcriptase Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
A proprietary MMLV mutant with superior robustness and reliability in RT reactions.
Thermo Scientific™ DreamTaq™ Hot Start Green PCR Master Mix
Thermo Scientific DreamTaq Hot Start DNA Polymerase is the product of choice when you need consistently robust amplification with minimal optimization steps
Invitrogen™ Platinum™ SuperFi™ DNA Polymerase Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Amplify even difficult and GC-rich amplicons with superior accuracy (>100X Taq) and minimaloptimization steps
Thermo Scientific™ phi29 DNA Polymerase Available on GSA/VA Contract for Federal Government customers only.
Perform highly efficient isothermal DNA amplification with phi29 DNA Polymerase, a highly processive polymerase with strong strand displacement activity.
Thermo Scientific™ DreamTaq Green DNA Polymerase Available on GSA/VA Contract for Federal Government customers only.
Save time in standard PCR applications with this enhanced Taq DNA polymerase, supplied with colored buffer enabling direct gel loading.
Thermo Scientific™ Taq DNA Polymerase, Recombinant Available on GSA/VA Contract for Federal Government customers only.
Optimize routine PCR with this highly thermostable DNA polymerase.
Invitrogen™ Platinum™ SuperFi™ PCR Master Mix Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Amplify even difficult and GC-rich amplicons with superior accuracy (>100X Taq) and minimaloptimization steps
Thermo Scientific™ DreamTaq DNA Polymerase Available on GSA/VA Contract for Federal Government customers only.
Get higher sensitivity, longer PCR products and higher yields in all standard PCR applications with this enhanced Taq DNA polymerase.
Promega™ GoTaq™ Flexi DNA Polymerase
Combines robust amplification, streamlined downstream analysis, and the flexibility to optimize Mg2+ concentration
Thermo Scientific™ Maxima Reverse Transcriptase Available on GSA/VA Contract for Federal Government customers only.
Increase synthesis rates with Thermo Scientific™ Maxima™ Reverse Transciptase, an advanced enzyme developed through in vitro evolution of M-MuLV RT.
Promega™ M-MLV Reverse Transcriptase
RNA-dependent DNA polymerase can be used in primer extension or cDNA synthesis with >5kb mRNA templates
Promega™ GoTaq™ G2 Green Master Mix
Designed for enhanced amplification
Promega™ AMV Reverse Transcriptase
Catalyzes the polymerization of DNA using template DNA, RNA, or RNA:DNA hybrids
Fisher BioReagents™ Taq DNA Polymerase Available on GSA/VA Contract for Federal Government customers only.
Allows up to 100 cycles of amplification without loss of activity
Thermo Scientific™ dsDNase Available on GSA/VA Contract for Federal Government customers only.
dsDNase is a heat inactivatable recombinant shrimp DNase used to remove genomic DNA contamination of RNA samples before reverse transcription.
Thermo Scientific™ DreamTaq Hot Start PCR Master Mix
Thermo Scientific DreamTaq Hot Start DNA Polymerase is the product of choice when you need consistently robust amplification with minimal optimization steps
Thermo Scientific™ RevertAid™ Reverse Transcriptase Available on GSA/VA Contract for Federal Government customers only.
Perform efficient synthesis of full-length first strand cDNA up to 13 kb with this genetically modified M-MuLV RT for routine reverse transcription.
Promega™ GoScript™ Reverse Transcriptase
Delivers robust, reliable cDNA synthesis of a full range of rare and abundant transcripts, even in the presence of inhibitors
Thermo Scientific™ DreamTaq Green DNA Polymerase (5 U/µL)
Thermo Scientific DreamTaq Green DNA Polymerase is a combination of DreamTaq DNA Polymerase and 10X DreamTaq Green Buffer.
Thermo Scientific™ DreamTaq™ Hot Start DNA Polymerase
DNA Polymerase is the product of choice for consistently robust amplification with minimal optimization steps
Thermo Scientific™ T4 DNA Polymerase Available on GSA/VA Contract for Federal Government customers only.
Ensure accuracy in DNA synthesis with template-dependent T4 DNA Polymerase that catalyzes the 5'-3' synthesis from primed single-stranded DNA.
Thermo Scientific™ DreamTaq™ Hot Start Green DNA Polymerase
Thermo Scientific DreamTaq Hot Start DNA Polymerase is the product of choice when you need consistently robust amplification with minimal optimization steps
Thermo Scientific™ Maxima H Minus Reverse Transcriptase Promotion
A highly thermostable, high processivity reverse transcriptase lacking RNase H activity
Thermo Scientific™ dsDNase: PROMO
dsDNase is a heat inactivatable recombinant shrimp DNase used to remove genomic DNA contamination of RNA samples before reverse transcription.
Thermo Scientific™ Maxima Reverse Transcriptase (200 U/µL)
Thermo Scientific Maxima Reverse Transcriptase (RT) was developed through in vitro evolution of M-MuLV RT
Thermo Scientific™ RevertAid H Minus Reverse Transcriptase Available on GSA/VA Contract for Federal Government customers only.
A recombinant M-MuLV RT lacking RNase H activity for high yield cDNA synthesis.
  spinner