qPCR Assays
Applied Biosystems™ TaqMan™ Array Human Hypoxia Non-distribution product as customer accommodation.
TaqMan™ Array Human Hypoxia 96-well Plate contains 44 assays to hypoxia signaling associated genes and 4 assays to candidate endogenous control genes.
Applied Biosystems™ Protein Thermal Shift™ Dye Kit Non-distribution product as customer accommodation.
This solution significantly reduces the time and cost associated with screening samples relative to currently available methods.
Applied Biosystems™ TaqMan™ Array Human NFkB Pathway 96-well Plate Non-distribution product as customer accommodation.
Easy to use - Since the primer/probe is supplied in a dry state, Just add the master mix and sample.
Applied Biosystems™ Protein Thermal Shift™ Starter Kit Non-distribution product as customer accommodation.
This solution significantly reduces the time and cost associated with screening samples relative to currently available methods.
Applied Biosystems™ TaqMan™ Array Human Immune Response Non-distribution product as customer accommodation.
TaqMan™ Array Human Immune Response 96-well Plate contains 92 assays to immune response associated genes and 4 assays to candidate endogenous control genes.
Applied Biosystems™ TaqMan™ Array Human Endogenous Control Non-distribution product as customer accommodation.
TaqMan™ Express Endogenous Control Plates use TaqMan™ probe-based chemistry and are designed for use on the suite of Applied Biosystems™ Real-Time PCR Systems.
Applied Biosystems™ TaqMan™ Array Human Inflammation Non-distribution product as customer accommodation.
The panel of assays in this plate targets genes involved in a broad range of inflammatory diseases.
Invitrogen™ Platinum™ Quantitative PCR SuperMix-UDG With ROX Non-distribution product as customer accommodation.
qPCR SuperMixes with ROX for ABI instruments are ready-to-use reaction mixes for high-specificity, real-time amplification of cDNA, genomic DNA or plasmid DNA.
Applied Biosystems™ TaqMan™ Array Human Chemokines Non-distribution product as customer accommodation.
TaqMan™ Array Plates contain the same high quality TaqMan™ Gene Expression Assays that have been commercially available for years in a convenient, dried, 96-well format.
EMD Millipore™ Novagen™ NovaQUANT™ Coli qPCR Kit
For accurate detection of trace amounts of bacterial DNA, typically in low femtogram range
Invitrogen™ Platinum™ Quantitative PCR SuperMix-UDG Non-distribution product as customer accommodation.
Is a ready-to-use mixture for high-specificity, real-time quantitative DNA amplification, containing UDG to prevent carryover contamination (1,2).
Invitrogen™ ROX Reference Dye Non-distribution product as customer accommodation.
ROX Reference Dye is designed to normalize the fluorescent reporter signal in real-time quantitative PCR or RT-PCR.
Applied Biosystems™ TaqMan™ Array Human DNA Methylation And Transcriptional Repression 96-well Plate Non-distribution product as customer accommodation.
Easy to use - Since the primer/probe is supplied in a dry state, Just add the master mix and sample.
Applied Biosystems™ TaqMan™ Array Human Apoptosis Non-distribution product as customer accommodation.
TaqMan™ Array Human Apoptosis 96-well Plate contains 92 assays to apoptosis associated genes and 4 assays to candidate endogenous control genes.
  spinner