RT-PCR Kits
Invitrogen™ SuperScript™ III First-Strand Synthesis System Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Optimized to synthesize first-strand cDNA from purified poly(A)+ or total RNA or RT-PCR
Applied Biosystems™ High-Capacity cDNA Reverse Transcription Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Delivers extremely high-quality, single-stranded cDNA from 0.02 to 2μg total RNA
Invitrogen™ SuperScript™ III Reverse Transcriptase Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Proprietary mutant of SuperScript II RT
Applied Biosystems™ TaqMan™ High-Capacity RNA-to-cDNA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Designed for optimum performance with TaqMan Gene Expression Master Mix, Power SYBR™ Green PCR Master Mix, and other PCR enzymes
Invitrogen™ SuperScript™ II Reverse Transcriptase Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
An improved version of SuperScript RT
Invitrogen™ SuperScript™ VILO™ cDNA Synthesis Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Provides the high-temperature capability of SuperScript III Reverse Transcriptase (RT) in an optimized format for generating first-strand cDNA for use in real-time quantitative RT-PCR (qRT-PCR)
Invitrogen™ SuperScript™ III First-Strand Synthesis SuperMix Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Optimized SuperMix formulation for first-strand cDNA synthesis from purified poly(A)+ or total RNA
Invitrogen™ SuperScript™ IV VILO™ Master Mix Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
SuperScript™ IV VILO™ Master Mix is a reaction master mix designed for fast, sensitive, and reproducible cDNA synthesis in RT-qPCR applications.
Applied Biosystems™ High-Capacity cDNA Reverse Transcription Kit with RNase Inhibitor Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Delivers extremely high-quality, single-stranded cDNA from 0.02 to 2μg total RNA
Invitrogen™ SuperScript™ III First-Strand Synthesis SuperMix for qRT-PCR Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Provides the high-temperature capability of SuperScript III Reverse Transcriptase in an optimized SuperMix format for synthesis of first-strand cDNA for use in real-time quantitative RT-PCR (qRT-PCR)
Invitrogen™ Platinum™ SuperScript™ III One-Step RT-PCR System with Platinum Taq DNA Polymerase Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Designed for sensitive, reproducible, end-point detection and analysis of RNA molecules by RT-PCR
Invitrogen™ SuperScript™ First-Strand Synthesis System for RT-PCR Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Optimized to synthesize first-strand cDNA from purified poly(A)+ or total RNA
Applied Biosystems™ TaqMan™ Reverse Transcription Reagents Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
For use with reverse transcriptase-PCR (RT-PCR)
Invitrogen™ MultiScribe™ Reverse Transcriptase Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
A recombinant moloney murine leukemia virus (rMoMuLV) reverse transcriptase that has been optimized for TaqMan-based assays
Invitrogen™ M-MLV Reverse Transcriptase Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Recombinant DNA polymerase that synthesizes a complementary DNA strand from single-stranded RNA, DNA, or an RNA:DNA hybrid
Invitrogen™ Platinum™ SuperScript™ One-Step RT-PCR System with Platinum Taq DNA Polymerase Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Convenient 2X format allows further addition of template and primer at any desired concentration
Biocartis Idylla™ Molecular Diagnostics Single-Use Cartridges
Idylla™ Single-Use Cartridges perform all processing steps in one cartridge.
Invitrogen™ RNase T1 (1U/μL) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Optimized for researchers performing RNA structure, RNA sequencing, protein footprinting, and boundary experiments. Invitrogen™ Ambion™ RNase T1 (1U/μL) includes RNase T1, 10X RNA Structure Buffer, 1X Alkaline Hydrolysis Buffer, Yeast RNA, Precipitation/Inactivation Buffer and Gel Loading Buffer II.
Invitrogen™ T7 RNA Polymerase-Plus Enzyme Mix (200U/μL) Non-distribution product as customer accommodation.
High-purity enzyme that's a mix of cloned RNA polymerase and RNase inhibitor
Promega GenePrint 10 System, 50 reactions
The GenePrint 10 System allows co-amplification and three-color detection of nine human loci, including the ASN-0002 loci (TH01, TPOX, vWA, Amelogenin, CSF1PO, D16S539, D7S820, D13S317 and D5S818) as well as D21S11.
Invitrogen™ T4 RNA Ligase (cloned) 5U/μL Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Catalyzes formation of a phosphodiester linkage between a 5'-phosphoryl-terminated nucleic acid donor and a 3'-hydroxyl-terminated nucleic acid acceptor
Invitrogen™ SP6 RNA Polymerase (cloned) 20U/μL Non-distribution product as customer accommodation.
A cloned high-purity RNA polymerase with a high specificity for its promoter
Invitrogen™ Cells-to-CT™ Power S 1-Step Power SYBR Green Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Makes it possible to perform expression analysis directly from cultured cells without RNA purification
Invitrogen™ Molecular Probes™ Amplex™ EnzChek™ Microplate Lysis Reagent Non-distribution product as customer accommodation.
A buffered nonionic detergent solution for the lysis of adherent or nonadherent cultured cells under non-denaturing conditions
Invitrogen™ Molecular Probes™ BackDrop™ Background Suppressor for Microplate Assays Non-distribution product as customer accommodation.
Designed to effectively suppress background fluorescence in live cell assays
Invitrogen™ Poly(A) Polymerase (cloned) 2U/μL Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Catalyzes the addition of adenosine to the 3' end of RNA in a sequence-independent fashion
  spinner