RT-qPCR Kits
Invitrogen™ SuperScript™ III Platinum™ One-Step qRT-PCR Kit Non-distribution product as customer accommodation.
Combine the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression in a convenient one-step format.
Applied Biosystems™ Power SYBR™ Green RNA-to-CT™ One-Step Kit Non-distribution product as customer accommodation.
Kit has been formulated to ensure maximum sensitivity and reliability. Get sensitivity and specificity in an easy-to-use one-step qRT-PCR reaction.
Applied Biosystems™ GeneScan™ 1200 LIZ™ Dye Size Standard
GeneScan 1200 LIZ Size Standard is designed for sizing DNA fragment in the 20-1200 nucleotides range and provides 68 single-stranded labeled fragment.
Invitrogen™ SuperScript™ III Platinum™ SYBR™ Green One-Step qPCR Kit With ROX Non-distribution product as customer accommodation.
Combine a powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression.
Applied Biosystems™ GeneScan™ 500 ROX™ dye Size Standard
GeneScan™ 500 ROX™ Size Standard is an internal lane size standard developed for use with the Applied Biosystems fluorescence-based DNA electrophoresis systems.
Invitrogen™ SuperScript™ III Platinum™ SYBR™ Green One-Step qRT-PCR Kit Non-distribution product as customer accommodation.
Combine the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression in a convenient one-step format.
Invitrogen™ SuperScript™ III Platinum™ One-Step qRT-PCR Kit With ROX Non-distribution product as customer accommodation.
One-Step qRT-PCR Kits with ROX combine the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression.
Invitrogen™ Platinum™ Quantitative RT-PCR ThermoScript™ One-Step System Non-distribution product as customer accommodation.
Platinum Quantitative RT-PCR ThermoScript One-Step System is for real-time quantitation of RNA molecules from total or poly(A)+ RNA in a single step.
Promega™ Plexor™ Two-Step RT-qPCR System
Less labor, less time and less cost per assay. Measure controls and targets at the same time in the same well. Each target is measured directly during the amplification process and not through a secondary reaction.
Applied Biosystems™ GeneScan™ 350 TAMRA™ Dye Size Standard
GeneScan 350 TAMRA Size Standard is designed for sizing DNA fragment in the 35-350 nucleotides range and provides 12 single-stranded labeled fragment.
Invitrogen™ SuperScript™ VILO™ Master Mix Available on GSA/VA Contract for Federal Government customers only.
SuperScript™ VILO™ Master Mix
6-Tamra NHS/DMSO
The single-isomer preparations of TAMRA are most important for high-resolution techniques such as DNA sequencing and capillary electrophoresis
Applied Biosystems™ GeneScan™ 500XL ROX™ dye Size Standard
GeneScan 500 ROX Size Standard is designed for sizing DNA fragments in the 35-500 nucleotides range and provides 16 single-stranded labeled fragment.
Applied Biosystems™ GeneScan™ 400HD ROX™ dye Size Standard
GeneScan 400 HD ROX Size Standard is designed for sizing DNA fragments in the 50-400 nucleotides range and provides 21 labeled fragments.
Applied Biosystems™ GeneScan™ 350 ROX™ dye Size Standard
GeneScan™350 ROX™ Size Standard is an internal lane size standard developed for use with the Applied Biosystems fluorescence based DNA electrophoresis systems.
Applied Biosystems™ GeneScan™ 600 LIZ™ Size Standard v2.0 Dx
Use this size standard for fragments between 20 and 600 bp. The standard contains 36 LIZ dye-labeled, single-stranded DNA fragments.
Applied Biosystems™ GeneScan™ 500XL TAMRA™ dye Size Standard
Designed for sizing DNA fragments in the 35-500 nucleotides range and provides 16 single-stranded labeled fragment.
Applied Biosystems™ DS-33 GeneScan Installation Kit with GeneScan 600 LIZ
GeneScan™ -120 LIZ® internal Size Standard is developed for use with the Applied Biosystems fluorescence based DNA electrophoresis systems. The use of internal size standard enables automated data analysis and is also essential for achieving high run to run precision in sizing DNA fragments.
Promega™ Plexor™ One-Step RT-qPCR System
Less labor, less time and less cost per assay. Measure controls and targets at the same time in the same well. Each target is measured directly during the amplification process and not through a secondary reaction.
Applied Biosystems™ AFLP Amplification Core Mix Module
The AFLP EcoRI and MseI Ligation/Amplification Modules provide sufficient reagents to modify and do preselective amplifications on 100 individual DNA samples (10 ng each).
  spinner