Antibody and Dye Based Assays
Invitrogen™ Molecular Probes™ Alexa Fluor™ 488 Phalloidin Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
High-affinity filamentous actin (F-actin) probe conjugated to bright, photostable, green-fluorescent Alexa Fluor 488 dye
R&D Systems™ Proteome Profiler Mouse Cytokine Array Kit, Panel A Available on GSA/VA Contract for Federal Government customers only.
For the parallel determination of the relative levels of selected mouse cytokines and chemokines
Invitrogen™ Molecular Probes™ Alexa Fluor™ 647 Phalloidin Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
High-affinity F-actin probe conjugated to bright, photostable, far-red fluorescent Alexa Fluor 647 dye
R&D Systems™ Proteome Profiler Human Phospho-Kinase Array Kit Available on GSA/VA Contract for Federal Government customers only.
Rapid, sensitive and economical tool to simultaneously detect the relative site-specific phosphorylation of 43 kinases and 2 related total proteins
Invitrogen™ Molecular Probes™ SYTOX™ Green Nucleic Acid Stain Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Useful indicator of dead cells within a population
eBioscience™ Fixable Viability Dye eFluor™ 780 Non-distribution product as customer accommodation.
Can be used to irreversibly label dead cells prior to cryopreservation, fixation and/or permeabilization procedures. Unlike 7-AAD and PI, cells labeled with Fixable Viability Dyes can be washed, fixed, permabilized and stained for intracellular antigens without loss of staining intensity.
Invitrogen™ Molecular Probes™ Alexa Fluor™ 568 Phalloidin Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
High-affinity F-actin probe conjugated to Alexa Fluor 568 dye
R&D Systems™ Proteome Profiler Human XL Cytokine Array Kit Available on GSA/VA Contract for Federal Government customers only.
Membrane-based antibody array for the parallel determination of the relative levels of selected human cytokines and chemokines
Invitrogen™ Molecular Probes™ MitoTracker™ Red CMXRos Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
For use with microarray, flow cytometry and fluorescence microscopy
R&D Systems™ Proteome Profiler Human Phospho-RTK Array Kit Available on GSA/VA Contract for Federal Government customers only.
For the parallel determination of the relative level of tyrosine phosphorylation of human receptor tyrosine kinases (RTKs)
Invitrogen™ Molecular Probes™ TO-PRO™-3 Iodide (642/661) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Useful as a nuclear counterstain and dead cell indicator
R&D Systems™ Proteome Profiler Human Angiogenesis Array Kit Available on GSA/VA Contract for Federal Government customers only.
Rapid, sensitive and economical tool to simultaneously detect the relative levels of 55 angiogenesis-related proteins in a single sample
Invitrogen™ Molecular Probes™ Alexa Fluor™ 594 Phalloidin Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
High-affinity F-actin probe conjugated to bright, photostable, red fluorescent Alexa Fluor 594 dye
Invitrogen™ Molecular Probes™ Concanavalin A, Alexa Fluor™ 488 Conjugate Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Concanavalin A (Con A) is widely used lectins in cell biology
Invitrogen™ Molecular Probes™ Rhodamine Phalloidin Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
High-affinity F-actin probe conjugated to red-orange fluorescent dye, tetramethylrhodamine (TRITC)
Invitrogen™ Molecular Probes™ LysoTracker™ Red DND-99 Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Red-fluorescent dye for labeling and tracking acidic organelles in live cells
  spinner