Antibody Production and Purification
Thermo Scientific™ Maleimide-Activated Blue Carrier™ Protein
Make hapten-carrier immunogens comprising cysteine-containing peptides and other thiolated antigens with sulfhydryl-reactive immunogenic hemocyanin (CCH).
Thermo Scientific™ Pierce™ Binding and Elution Buffers for IgG Purification Available on GSA/VA Contract for Federal Government customers only.
Ensure high-yield, nondenaturing antibody purification using IgG affinity purification supports with these gentle buffer solutions.
Thermo Scientific™ Pierce™ Fab Preparation Kits Available on GSA/VA Contract for Federal Government customers only.
Digest human or mouse IgG antibodies with immobilized papain protease and purify Fab fragments using Protein A agarose.
Thermo Scientific™ Imject™ Mariculture KLH and EDC Conjugation Kit
Prepare immunogens (hapten-carrier conjugates) with your antigen of choice and mariculture KLH using different conjugation chemistries.
Thermo Scientific™ Easy-Titer™ Antibody Assay Kits
Determine antibody concentration in serum and culture supernatant with kits specific for mouse, rabbit, or human IgG, and human IgM.
Thermo Scientific™ Melon Gel™ IgG Purification Resins and Kits Available on GSA/VA Contract for Federal Government customers only.
Rapidly purify human, mouse and rabbit IgG from serum and other sources with this simpler alternative to traditional Protein A and Protein G methods.
Thermo Scientific™ Imject™ Maleimide-Activated mcKLH and Kit Available on GSA/VA Contract for Federal Government customers only.
Perform hapten-carrier conjugation with cysteine-containing peptides and other thiolated antigens with this sulfhydryl-reactive mariculture KLH.
Thermo Scientific™ Imject™ Maleimide-Activated BSA and Kit
Prepare hapten-carrier conjugates with cysteine-containing peptides and other thiolated antigens using this sulfhydryl-reactive bovine serum albumin.
Thermo Scientific™ Blue Carrier™ Protein and EDC Conjugation Kit
Easily prepare effective immunogens with the highly-soluble, mollusk-derived hemocyanin and complete hapten-carrier conjugation kit.
Thermo Scientific™ Imject™ Maleimide-Activated Ovalbumin
Prepare hapten-carrier conjugates to generate antibodies against cysteine-containing peptides and other thiolated antigens using this sulfhydryl-reactive ovalbumin protein.
Thermo Scientific™ Pierce™ Mouse IgG1 Fab and F(ab')2 Preparation Kits Available on GSA/VA Contract for Federal Government customers only.
Efficiently digest mouse IgG1 antibodies into Fab or F(ab')2 fragments with immobilized ficin protease.
Thermo Scientific™ Imject™ Ovalbumin Carrier Protein
Prepare hapten-carrier conjugates to generate antibodies using this conveniently formulated, purified ovalbumin (OVA) protein.
Thermo Scientific™ Pierce™ Rapid Mouse Antibody Isotyping Kits
Identify mouse monoclonal antibody class and subclass in 5 minutes with these single-use, cassette-based lateral-flow assays.
Novus Biologicals™ Antibody Concentration and Clean Up Antibody Purification Kit
Allows quick and easy concentration of antibodies and proteins
Thermo Scientific™ Imject™ Freund's Adjuvants
Stimulate a nonspecific immune response using complete (CFA) or incomplete (IFA) formulations of the popular Freund's adjuvant; offered in 10mL ampules.
Thermo Scientific™ Pierce™ F(ab')2 Preparation Kits
Selectively digest whole human and other IgG antibodies with immobilized pepsin protease to make F(ab')2 fragments that retain antigen binding activity.
Thermo Scientific™ Pierce™ Thiophilic Adsorbent Resin and Kit
Preferentially bind and purify immunoglobulins from various sources including mouse, rat, rabbit, goat and human with this non-proteinaceous immunosorbent resin.
Thermo Scientific™ Imject™ BSA Protein and EDC Conjugation Kit
Easily prepare immunogens from carboxyl-containing peptide antigens with bovine serum albumin (BSA) and different conjugation chemistries.
Thermo Scientific™ Pierce™ Rapid ELISA Mouse mAb Isotyping Kit
Isotype mouse monoclonal antibody using an antigen-independent, microplate-based method that enables multiple samples to be tested in about one hour.
Thermo Scientific™ Imject™ Alum Adjuvant Available on GSA/VA Contract for Federal Government customers only.
Stimulate a nonspecific immune response for mixed antigens (prepared immunogens) using this formulation of aluminum hydroxide and magnesium hydroxide.
BD Difco™ Adjuvants
Used to prepare antigen-adjuvant emulsions. BD™ Difco™ Adjuvants are used in immunological studies with laboratory animals. (Not intended for human or therapeutic use.)
Thermo Scientific™ Pierce™ Saturated Ammonium Sulfate Solution
Fractionate immunoglobulin, antibody, or other proteins by selective precipitation, also called salting out.
Thermo Scientific™ Melon Gel™ Monoclonal IgG Purification Kit
Purify mouse monoclonal antibodies from serum, ascites or hybridoma cell culture supernatant with this high-yield, mild monoclonal IgG purification kit.
GE Healthcare GammaBind Plus Sepharose Available on GSA/VA Contract for Federal Government customers only.
Binds to all IgG subclasses. GE Healthcare GammaBind Plus Sepharose is used for lab-scale affinity purification of IgG from a large variety of species and has better binding to rat and mouse IgG subclasses compared to standard GammaBind Sepharose.
Thermo Scientific™ Pierce™ IgM Purification Kit Available on GSA/VA Contract for Federal Government customers only.
Easily purify IgM from mouse ascites fluid using immobilized mannan binding protein, which binds to IgM in the kit-supplied buffers.
HiTrap™ MabSelect™ SuRe Prepacked Columns Available on GSA/VA Contract for Federal Government customers only.
Use these columns in small-scale MAb purification in a continuous bioprocess. GE Healthcare Life Sciences™ HiTrap™ MabSelect™  SuRe (Superior Resistance) Prepacked Columns for fast purification of monoclonal antibodies (Mabs) from large sample volumes
Thermo Scientific™ Pierce™ Jacalin Agarose IgA Purification Resin
Purify human IgA from serum or colostrum or efficiently remove contaminating IgA from IgG samples with this specific affinity resin.
  spinner