Adjuvants for Immunization
BD Difco™ Adjuvants
Used to prepare antigen-adjuvant emulsions. BD™ Difco™ Adjuvants are used in immunological studies with laboratory animals. (Not intended for human or therapeutic use.)
Thermo Scientific™ Imject™ Freund's Adjuvants
Stimulate a nonspecific immune response using complete (CFA) or incomplete (IFA) formulations of the popular Freund's adjuvant; offered in 10mL ampules.
Thermo Scientific™ Imject™ Alum Adjuvant
Stimulate a nonspecific immune response for mixed antigens (prepared immunogens) using this formulation of aluminum hydroxide and magnesium hydroxide.
  spinner