Antibody Fragmentation Kits
Thermo Scientific™ Pierce™ Fab Preparation Kits Available on GSA/VA Contract for Federal Government customers only.
Digest human or mouse IgG antibodies with immobilized papain protease and purify Fab fragments using Protein A agarose.
Thermo Scientific™ Pierce™ Mouse IgG1 Fab and F(ab')2 Preparation Kits Available on GSA/VA Contract for Federal Government customers only.
Efficiently digest mouse IgG1 antibodies into Fab or F(ab')2 fragments with immobilized ficin protease.
Thermo Scientific™ Pierce™ F(ab')2 Preparation Kits
Selectively digest whole human and other IgG antibodies with immobilized pepsin protease to make F(ab')2 fragments that retain antigen binding activity.
  spinner