Antibody Isotyping and Characterization
Thermo Scientific™ Pierce™ Rapid Mouse Antibody Isotyping Kits
Identify mouse monoclonal antibody class and subclass in 5 minutes with these single-use, cassette-based lateral-flow assays.
Thermo Scientific™ Pierce™ Rapid ELISA Mouse mAb Isotyping Kit
Isotype mouse monoclonal antibody using an antigen-independent, microplate-based method that enables multiple samples to be tested in about one hour.
Thermo Scientific™ Easy-Titer™ Antibody Assay Kits
Determine antibody concentration in serum and culture supernatant with kits specific for mouse, rabbit, or human IgG, and human IgM.
MilliporeSigma™ Novagen™ FastPlax™ Titer Kit
Designed for determination of baculovirus titers.
  spinner