BSA Carrier Protein Immunogens
Thermo Scientific™ Imject™ BSA Protein and EDC Conjugation Kit
Easily prepare immunogens from carboxyl-containing peptide antigens with bovine serum albumin (BSA) and different conjugation chemistries.
Thermo Scientific™ Imject™ Maleimide-Activated BSA and Kit
Prepare hapten-carrier conjugates with cysteine-containing peptides and other thiolated antigens using this sulfhydryl-reactive bovine serum albumin.
EMD Millipore™ Calbiochem™ Advanced Glycation End product-BSA Available on GSA/VA Contract for Federal Government customers only.
Prepared by reacting BSA with glycoaldehyde under sterile conditions.
EMD Millipore™ Calbiochem™ BSA, Fraction V, Fatty Acid-Poor, Endotoxin-Free Available on GSA/VA Contract for Federal Government customers only.
Designed for use in cell culture systems, animal perfusion studies.
EMD Millipore™ Calbiochem™ BSA, Fraction V, Low Heavy Metals Available on GSA/VA Contract for Federal Government customers only.
Prepared by heat shock treatment procedure.
EMD Millipore™ Calbiochem™ Bovine Serum Albumin, Fraction V (Fatty Acid, Nuclease and Protease Free) Available on GSA/VA Contract for Federal Government customers only.
Designed for use in serological testing, RIA, hormone response studies.
EMD Millipore™ Calbiochem™ Prionex™Reagent Available on GSA/VA Contract for Federal Government customers only.
Used as alternative to bovine serum albumin.
EMD Millipore™ Calbiochem™ Bovine Serum Albumin, Fraction V (Modified Cohn) Available on GSA/VA Contract for Federal Government customers only.
Reflects endogenous lipids with native fatty acid profile.
  spinner