Buffers for Antibody Affinity Purification
Thermo Scientific™ Pierce™ Binding and Elution Buffers for IgG Purification Available on GSA/VA Contract for Federal Government customers only.
Ensure high-yield, nondenaturing antibody purification using IgG affinity purification supports with these gentle buffer solutions.
R&D Systems™ Reconstitution Buffer 4 (BSA/HCl)
0.1% Bovine Serum Albumin in 4 mM HCl
R&D Systems™ Reconstitution Buffer 6 (BSA/Acetic Acid)
0.1% Bovine Serum Albumin in 100 mM Acetic Acid
R&D Systems™ Reconstitution Buffer 7 (Deionized Water)
This product is intended as a companion product for reconstitution of selected lyophilized antibody or protein to maximize recovery and activity.
R&D Systems™ Reconstitution Buffer 2 (BSA/PBS)
0.1% Bovine Serum Albumin in PBS
Alfa Aesar™ Antibody & Antigen Purification Buffers
Buffers for immunoaffinity purification of antibodies
  spinner