KLH Carrier Protein Immunogens
Thermo Scientific™ Imject™ Maleimide-Activated mcKLH and Kit Available on GSA/VA Contract for Federal Government customers only.
Perform hapten-carrier conjugation with cysteine-containing peptides and other thiolated antigens with this sulfhydryl-reactive mariculture KLH.
Thermo Scientific™ Imject™ Mariculture KLH and EDC Conjugation Kit
Prepare immunogens (hapten-carrier conjugates) with your antigen of choice and mariculture KLH using different conjugation chemistries.
  spinner