Misc Antibody Purification Products
Novus Biologicals™ Antibody Concentration and Clean Up Antibody Purification Kit
Allows quick and easy concentration of antibodies and proteins
Thermo Scientific™ Pierce™ Thiophilic Adsorbent Resin and Kit
Preferentially bind and purify immunoglobulins from various sources including mouse, rat, rabbit, goat and human with this non-proteinaceous immunosorbent resin.
Thermo Scientific™ Pierce™ Saturated Ammonium Sulfate Solution
Fractionate immunoglobulin, antibody, or other proteins by selective precipitation, also called salting out.
Thermo Scientific™ Pierce™ IgM Purification Kit Available on GSA/VA Contract for Federal Government customers only.
Easily purify IgM from mouse ascites fluid using immobilized mannan binding protein, which binds to IgM in the kit-supplied buffers.
Thermo Scientific™ Pierce™ Jacalin Agarose IgA Purification Resin
Purify human IgA from serum or colostrum or efficiently remove contaminating IgA from IgG samples with this specific affinity resin.
GE Healthcare HiTrap™ IgY Purification Column Available on GSA/VA Contract for Federal Government customers only.
Developed for fast, easy purification of IgY from chicken egg yolk
Novus Biologicals™ AbSelect Antibody Purification Kit
Fast and simple method to purify antibodies from ascites fluid and serum
Thermo Scientific™ Pierce™ Chicken IgY Purification Kits and Reagents
Purify chicken antibodies from hen egg yolks with a unique delipidation reagent and precipitation reagent to selectively isolate the IgY fraction.
GE Healthcare HiTrap™ IgM Purification HP Column Available on GSA/VA Contract for Federal Government customers only.
Optimized for fast, efficient purification of monoclonal IgM from hybridoma cell culture
Novus Biologicals™ AbSelect G Serum System Antibody Purification Kit
For purification of antibodies from crude samples such as ascites fluid or immune serum
Novus Biologicals™ AbSelect Serum Antibody Purification Kit
Fast and simple method to purify antibodies from ascites fluid and serum
Novus Biologicals™ Eight - Tube Magnetic Stand
For use with AbPure Magnetic Antibody Purification Kit
Novus Biologicals™ AbSelect G System Antibody Purification Kit
For purification of antibodies from crude samples such as ascites fluid or immune serum and removal of contaminants
Novus Biologicals™ AbSelect Mouse System Antibody Purification Kit
For purification of mouse antibodies from crude samples such as ascites fluid
Thermo Scientific™ Pierce™ Antibody Purification Collection Plates
Collection plates and balance plates for use with 96-well filter spin plates
Novus Biologicals™ AbSelect Mouse TCS System Antibody Purification Kit
For purification of mouse IgG fractions from hybridoma supernatants
GE Healthcare HiScreen MabSelect Xtra Available on GSA/VA Contract for Federal Government customers only.
Prepacked with MabSelect Xtra. GE Healthcare HiScreen MabSelect Xtra is part of the GE Healthcare process development platform. The columns are an excellent choice for method optimization and parameter screening for capture of monoclonal antibodies (MAbs).
EMD Millipore™ Novagen™ His·Tag Antibody Plate Available on GSA/VA Contract for Federal Government customers only.
ELISA-compatible plate containing immobilized His·Tag monoclonal antibodies
  spinner