Cypridina Luciferase Vectors and Assays
Thermo Scientific™ Pierce™ Cypridina Luciferase Flash Assay Kit
Assay regulatory element transcriptional activity in mammalian cell culture media and whole cell lysate with this highly sensitive flash-based luciferase kit.
Thermo Scientific™ Cypridina Luciferase Reporter Assay Vectors
Drive Cypridina luciferase expression as a reporter for gene regulation studies using vectors with a multiple cloning site (MCS) or control promoter (CMV or TK).
Thermo Scientific™ Pierce™ Cypridina-Firefly Luciferase Dual Assay Kit
Simultaneously detect intracellular Cypridina and Red Firefly luciferase activity in mammalian whole cell lysates.
Thermo Scientific™ Pierce™ Cypridina Luciferase Glow Assay Kit
Obtain bright and stable bioluminescence signal (half-life greater than 1 hour) in the presence of Cypridina luciferase reporters.
  spinner