Gaussia-Dura Luciferase Vectors and Assays
Thermo Scientific™ Pierce™ Gaussia Luciferase Glow Assay Kit Available on GSA/VA Contract for Federal Government customers only.
Obtain bright and stable bioluminescence signal (half-life greater than 1 hour) in the presence of Gaussia luciferase reporters
Thermo Scientific™ Gaussia-Dura Luciferase Reporter Assay Vectors
Drive mutated Gaussia luciferase expression with vectors containing a multiple cloning site (MCS) or control promoter (CMV or TK) for extended signal output.
  spinner