Class Specific Protein Assay Kits
Promega™ ROS-Glo™ H2O2 Assay
Measures the level of hydrogen peroxide (H2O2), a reactive oxygen species (ROS), directly in cell culture or in defined enzyme reactions. Light signal that is proportional to the level of H2O2 present in the sample.
Thermo Scientific™ Pierce™ Protease Assay Kits
Determine total protease activity in biological samples via fluorescent or colorimetric detection by measuring digestion of labeled casein substrate.
Thermo Scientific™ Pierce™ Glycoprotein Carbohydrate Estimation Kit
Measure protein glycosylation as the percent of purified protein mass compared to included glycoprotein standards; uses periodic acid Schiff-base reagents (PAS).
R&D Systems™ Malachite Green Phosphate Detection Kit Available on GSA/VA Contract for Federal Government customers only.
contains sufficient materials to run Sandwich ELISAs on at least 1 Kit
Promega™ DPPIV-Glo™ Protease Assay
Homogeneous, luminescent assay that measures dipeptidyl peptidase IV (DPPIV) activity
Abnova™ Thioredoxin Reductase Assay Kit Available on GSA/VA Contract for Federal Government customers only.
Thioredoxin Reductase Assay Kit is used for detecting TrxR activity.
Thermo Scientific™ Pierce™ Phosphoprotein Phosphate Estimation Kit
Classify and identify proteins as phosphorylated (serine and threonine) and obtain semi-quantitative assessment of the phosphorylation levels.
Boston Biochem™ MgATP Solution
General Ubiquitin Proteasome Pathway (UPP) Products
Boston Biochem™ 10X Energy Regeneration Solution
General Ubiquitin Proteasome Pathway (UPP) Products
Boston Biochem™ HSP Reaction Buffer
General Ubiquitin Proteasome Pathway (UPP) Products
Boston Biochem™ 10X E3 Ligase Reaction Buffer
General Ubiquitin Proteasome Pathway (UPP) Products
Boston Biochem™ 10X Ubiquitin Conjugation Reaction Buffer
Formulated to support the charging of Ubiquitin by E1 activating enzymes (the first reaction in the Ubiquitin conjugation cascade)
Boston Biochem™ 10X SUMO Conjugation Reaction Buffer
Contains the optimal formulation for the use in conjugation assays of the ubiquitin-like modifiers, SUMO1 (UL-712), SUMO2 (UL-752), and SUMO3 (UL-762), to protein substrates in vitro.
R&D Systems™ S-100 Fraction Conjugation Kit, HeLa Cells
For the controlled conjugation of exogenously added or endogenously contained substrate proteins through the ubiquitin proteasome system (UPS)
Boston Biochem™ 200X Conjugation Stop Buffer
General Ubiquitin Proteasome Pathway (UPP) Products
Boston Biochem™ 10X Ubiquitin Reconstitution Buffer
Formulated to provide the optimal formulation for the reconstitution and subsequent dilution of lyophilized Ubiquitin and Ubiquitin-like proteins
Boston Biochem™ 10X HSP60 Reaction Buffer
Formulated to support the in vitro enzymatic activity of Heat Shock Proteins, specifically HSP60 and HSP60/HSP10 complex
Boston Biochem™ AMC Standard
General Ubiquitin Proteasome Pathway (UPP) Products
Invitrogen™ KDalert™ GAPDH Assay Kit Non-distribution product as customer accommodation.
KDalert™ GAPDH Assay Kit
Boston Biochem™ 20X 20S Proteasome Reaction Buffer
Contains the optimal formulation for the quantitative analysis of constitutive 20S and immuno 20S proteasome activity
Boston Biochem™ 10X HSP90 Reaction Buffer
Formulated to support the in vitro enzymatic activity of Heat Shock Proteins, specifically HSP90 and the HSP90/p23 complex
Boston Biochem™ 100X 20S Proteasome Activation Solution
Contains the optimal formulation for the chemical activation of all isoforms of constitutive 20S and immuno 20S proteasome enzymes including human (E-360), rabbit (E-350), rat (E-375), and mouse.
Boston Biochem™ 10X NEDD8 Conjugation Reaction Buffer
Formulated to support in vitro conjugation assays with the Ubiquitin-like modifier NEDD8 (UL-812)
Invitrogen™ KDalert™ GAPDH Assay Kit with Manual Non-distribution product as customer accommodation.
KDalert™ GAPDH Assay Kit with Manual
Boston Biochem™ AFC Standard
General Ubiquitin Proteasome Pathway (UPP) Products
EMD Millipore™ Calbiochem™ CB-Protein Assay™ Kit
Used for estimation of protein concentration.
Zeptometrix Formamido Cytidine Dimer d(PfpC) Dinucleotide Standard Available on GSA/VA Contract for Federal Government customers only.
May be used as an internal standard for various quantitations
EMD Millipore™ Calbiochem™ Non-Interfering Protein Assay™ Kit
Overcomes interference of agents found in protein solutions.
Zeptometrix OXItek Total Glutathione Peroxidase Assay Kit Available on GSA/VA Contract for Federal Government customers only.
Provides a method of quantifying the activity of total glutathione peroxidase
  spinner