Carboxyl-to-Amine Crosslinkers
Thermo Scientific™ Pierce™ Sulfo-NHS Crosslinker Available on GSA/VA Contract for Federal Government customers only.
Prepare amine-reactive sulfo-NHS-ester crosslinkers or enhance carbodiimide (EDC) conjugation for peptides or proteins using N-hydroxysulfosuccinimide, CAS 106627-54-7.
Thermo Scientific™ Pierce™ EDC Crosslinker Available on GSA/VA Contract for Federal Government customers only.
Immobilize or conjugate proteins using 1-Ethyl-3-(3-dimethylaminopropyl) carbodiimide (CAS 25952-53-8), a water-soluble, carboxyl-to-amine crosslinker.
Thermo Scientific™ Pierce™ Premium Grade Sulfo-NHS
Optimize applications where product integrity and risk minimization are paramount with our highest quality formulation of this N-hydroxysuccinimide modification reagent.
Thermo Scientific™ Pierce™ NHS Crosslinker
Prepare amine-reactive NHS-ester crosslinkers or enhance carbodiimide (EDC) conjugation for peptides or proteins using N-hydroxysuccinimide, CAS 6066-82-6.
Thermo Scientific™ Pierce™ Premium Grade EDC
Optimize applications where product integrity and risk minimization are paramount with our highest quality formulation of this popular carbodiimide crosslinker.
Thermo Scientific Pierce™ EDC, No-Weigh™ Format Crosslinker
Thermo Scientific™ Pierce™ EDC is carboxyl- and amine-reactive zero-length crosslinker. EDC reacts with carboxyl group first and forms amine-reactive O-acylisourea intermediate that quickly reacts with amino group to form amide bond with release of isourea by-product.
  spinner