Cross Linking Reagents
Thermo Scientific™ EZ-Link™ Sulfo-NHS-LC-Desthiobiotin No-Weigh™ Format
Thermo Scientific™ EZ-Link™ Sulfo-NHS-LC-Desthiobiotin No-Weigh™ Format is a long-chain, amine-reactive labeling reagent whose biotin-like group is elutable from streptavidin, making it ideal for purified or cell surface protein labeling and purification.
EMD Millipore™ Calbiochem™ SMCC, Water-Soluble
Used for coupling enzymes to antibodies.
Thermo Scientific™ Pierce™ Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate) No-Weigh™ Format
Thermo Scientific™ Pierce™ Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate) No-Weigh™ Format is a water-soluble, amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium cyclohexane spacer arm.
Thermo Scientific™ EZ-Link™ Phosphine-PEG4-Desthiobiotin No-Weigh™ Format
Thermo Scientific™ EZ-Link™ Phosphine-PEG4-Desthiobiotin No-Weigh™ Format is a pegylated reagent whose biotin-like group is elutable from streptavidin, activated for chemoselective ligation and purification of azide targets tagged by metabolic labeling.
Thermo Scientific™ Pierce™ DSS (disuccinimidyl suberate) No-Weigh™ Format
Thermo Scientific™ Pierce™ DSS (disuccinimidyl suberate) No-Weigh™ Format is noncleavable and membrane permeable crosslinker that contains an amine-reactive N-hydroxysuccinimide (NHS) ester at each end of an 8-carbon spacer arm.
Thermo Scientific™ EZ-Link™ Maleimide-PEG2-Biotin No-Weigh™ Format
Thermo Scientific™ EZ-Link™ Maleimide-PEG2-Biotin No-Weigh™ Format is a mid-length, maleimide-activated, sulfhydryl-reactive biotinylation reagent that contains a 2-unit ethylene glycol in its spacer arm for increased water-solubility characteristics.
EMD Millipore™ Calbiochem™ SATA
Reacts with primary amines to add protected sulfhydryl groups.
Thermo Scientific™ Pierce™ Sulfo-NHS (N-hydroxysulfosuccinimide) No-Weigh™ Format
Thermo Scientific™ Pierce™ Sulfo-NHS (N-hydroxysulfosuccinimide) No-Weigh™ Format is a chemical modification reagent for converting carboxyl groups to amine-reactive NHS esters for bioconjugation, crosslinking, labeling and immobilization methods.
EMD Millipore™ Calbiochem™ SPDP Available on GSA/VA Contract for Federal Government customers only.
Used for introducing protected thiol groups.
  spinner