Imidoester Crosslinkers
Thermo Scientific™ Pierce™ DMP Crosslinker
Crosslink amines to amines with dimethyl pimelimidate (CAS 58537-94-3); composed of imidoester groups and a membrane-permeable, 7-atom spacer arm.
Thermo Scientific™ Pierce™ DTBP Crosslinker
Crosslink amines to amines with dithiobispropionimidate; composed of imidoester groups at either end of a reducible (cleavable) disulfide-containing spacer arm.
Thermo Scientific™ Pierce™ DMS Crosslinker
Crosslink amines to amines with dimethyl suberimidate (CAS 34490-86-3); composed of imidoester groups at either end of a membrane-permeable, 8-atom spacer arm.
Thermo Scientific™ Pierce™ DMA Crosslinker
Crosslink amines to amines with dimethyl adipimidate (CAS 14620-72-5); composed of imidoester groups and a 6-atom, membrane-permeable spacer arm.
  spinner