NHS-ester Crosslinkers
Thermo Scientific™ Pierce™ BS3 (Sulfo-DSS) Crosslinker Available on GSA/VA Contract for Federal Government customers only.
Crosslink amines to amines with BS3; composed of sulfo-NHS-ester groups separated by an 11.4A, water-soluble spacer arm that is useful for cell-surface protein crosslinking.
Thermo Scientific™ Pierce™ BS(PEG)n Crosslinkers Available on GSA/VA Contract for Federal Government customers only.
Crosslink amines to amines with these long (21.7A and 35.8A) crosslinkers composed of NHS-ester groups and 5-unit or 9-unit PEG spacer arms that enhance protein solubility.
Thermo Scientific™ Pierce™ DSG Crosslinker Available on GSA/VA Contract for Federal Government customers only.
Crosslink amines to amines with disuccinimidyl glutarate with NHS-ester groups at the ends of a 7.7A spacer arm; membrane-permeable for intracellular protein crosslinking.
Thermo Scientific™ Pierce™ DSP Crosslinker Available on GSA/VA Contract for Federal Government customers only.
Crosslink amines to amines with dithiobis succinimidyl propionate, composed of NHS-ester groups and disulfide-containing, 12.0A spacer arm and cleavable with reducing agents.
Thermo Scientific™ Pierce™ EGS Crosslinker Available on GSA/VA Contract for Federal Government customers only.
Crosslink amines to amines with this membrane permeable crosslinker composed of NHS-ester groups and a 16.1A hydroxylamine-cleavable spacer arm.
Thermo Scientific™ Pierce™ DTSSP Crosslinker Available on GSA/VA Contract for Federal Government customers only.
Crosslink amines to amines with Sulfo-DSP; composed of sulfo-NHS-ester groups and disulfide spacer arm (12.0A) that is cleavable with reducing agents.
Thermo Scientific™ Pierce™ Premium Grade DSP
Optimize applications where product integrity and risk minimization are paramount with our highest quality formulation of this popular amine-reactive, cleavable crosslinker.
Thermo Scientific Pierce™ DSP, No-Weigh™ Format Crosslinker
Thermo Scientific™ DSP (Lomant's Reagent) is water-insoluble, homo-bifunctional N-hydroxysuccimide ester (NHS ester) crosslinker that is thiol-cleavable, primary amine-reactive, and useful for many applications.
Thermo Scientific™ Pierce™ DSS Crosslinker
Crosslink amines to amines with disuccinimidyl suberate composed of NHS-ester groups with a membrane-permeable, mid-length spacer for intracellular protein crosslinking.
Thermo Scientific™ Pierce™ Sulfo-EGS Crosslinker
Crosslink amines to amines with a crosslinker composed of sulfo-NHS-ester groups and hydroxylamine-cleavable spacer arm; water-soluble for cell-surface protein crosslinking.
Thermo Scientific Pierce™ SM(PEG)2, No-Weigh™ Format Crosslinker
Thermo Scientific™ Pierce™ SM(PEG)2 is hetero-bifunctional crosslinker with N-hydroxysuccinimide (NHS) ester and maleimide groups that allow covalent conjugation of amine- and sulfhydryl-containing molecules.
Thermo Scientific™ Pierce™ DST Crosslinker
Crosslink amines to amines with disuccinimidyl tartrate, composed of NHS-ester groups and a diol-containing spacer arm (6.4A) that is periodate-cleavable.
Thermo Scientific Pierce™ Sulfo-SANPAH, No-Weigh™ Format Crosslinker
Thermo Scientific™ Pierce™ Sulfo-SANPAH is hetero-bifunctional crosslinker that contains amine-reactive N-hydroxysuccinimide (NHS) ester and photoactivatable nitrophenyl azide.
Thermo Scientific Pierce™ DSG, No-Weigh™ Format Crosslinker
Thermo Scientific™ Pierce™ DSG is water-insoluble, homo-bifunctional N-hydroxysuccinimide ester (NHS-ester) crosslinker often used for conjugating radiolabeled ligands to cell-surface receptors.
Thermo Scientific Pierce™ SM(PEG)12, No-Weigh™ Format Crosslinker
Thermo Scientific™ Pierce™ SM(PEG)12 is hetero-bifunctional crosslinker with N-hydroxysuccinimide (NHS) ester and maleimide groups that allow covalent conjugation of amine- and sulfhydryl-containing molecules.
Thermo Scientific Pierce™ SMCC, No-Weigh™ Format Crosslinker
Thermo Scientific™ Pierce™ SMCC is hetero-bifunctional crosslinker that contain N-hydroxysuccinimide (NHS) ester and maleimide groups that allow covalent conjugation of amine- and sulfhydryl-containing molecules.
Thermo Scientific™ Pierce™ BSOCOES Crosslinker
Crosslink amines to amines with this crosslinker composed of NHS-ester groups and a sulfone-containing spacer arm (13.0A) that is base-cleavable.
Thermo Scientific™ Pierce™ Premium Grade BS3
Crosslink proteins and other biomolecules in applications where product integrity and risk minimization are paramount with Thermo Scientific™ Pierce™ Premium Grade BS3, a high-quality formulation of this popular amine-reactive crosslinker.
Thermo Scientific™ Pierce™ TSAT Crosslinker
A 3-way, short-arm (4.2A) amine-to-amine crosslinker; tris (succinimidyl) aminotriacetate, composed of three NHS-ester groups on short spacer arms; membrane permeable.
  spinner