NHS-Haloacetyl Crosslinkers
Thermo Scientific™ Pierce™ SIA Crosslinker
Crosslink amines to sulfhydryls with this very short crosslinker composed of NHS-ester and iodoacetyl (haloacetyl) groups at ends of a 3-atom (1.5A) spacer arm.
Thermo Scientific™ Pierce™ Sulfo-SIAB Crosslinker
Crosslink amines to sulfhydryls with this mid-length, water-soluble crosslinker composed of sulfo-NHS-ester and iodoacetyl (haloacetyl) groups at ends of a 9-atom spacer arm.
Thermo Scientific™ Pierce™ SBAP Crosslinker
Crosslink amines to sulfhydryls with this short crosslinker composed of NHS-ester and bromoacetyl (haloacetyl) groups at ends of a 7-atom (6.2A) spacer arm.
Thermo Scientific™ Pierce™ SIAB Crosslinker
Crosslink amines to sulfhydryls with this mid-length crosslinker composed of NHS-ester and iodoacetyl (haloacetyl) groups at ends of a 9-atom (10.6A) aminobenzoate spacer arm.
  spinner