NHS-Maleimide Crosslinkers
Thermo Scientific™ Pierce™ SMCC Crosslinker
Crosslink amines to sulfhydryls with this mid-length crosslinker composed of NHS-ester and maleimide groups at ends of a stabilizing cyclohexane-containing, 8-atom spacer arm.
Thermo Scientific™ Pierce™ Sulfo-SMCC Crosslinker
Crosslink amines to sulfhydryls with this mid-length, water-soluble crosslinker made of NHS-ester and maleimide groups at ends of a cyclohexane-containing, 8-atom spacer arm.
Thermo Scientific™ Pierce™ MBS Crosslinker
Crosslink amines to sulfhydryls with this short crosslinker composed of NHS-ester and maleimide groups at ends of an aromatic, 6-atom (7.3A) spacer arm.
Thermo Scientific™ Pierce™ SM(PEG)n Crosslinkers Available on GSA/VA Contract for Federal Government customers only.
Crosslink amines to sulfhydryls with these crosslinkers containing NHS-ester and maleimide groups at ends of water-soluble polyethylene glycol spacer arms (17.6 to 95.2A).
Thermo Scientific™ Pierce™ GMBS Crosslinker
Crosslink amines to sulfhydryls with this short crosslinker composed of NHS-ester and maleimide groups at ends of an aliphatic, 6-atom (7.3A) spacer arm.
Thermo Scientific™ Pierce™ SMPB Crosslinker
Crosslink amines to sulfhydryls with this mid-length crosslinker composed of NHS-ester and maleimide groups at ends of an aromatic, 10-atom (11.6A) spacer arm.
Thermo Scientific™ Pierce™ LC-SMCC Crosslinker
Crosslink amines to sulfhydryls with this long crosslinker composed of NHS-ester and maleimide groups at ends of a stabilizing cyclohexane-containing, 15-atom spacer arm.
Thermo Scientific™ Pierce™ BMPS Crosslinker
Crosslink amines to sulfhydryls with this very short amine-to-sulfhydryl crosslinker composed of NHS-ester and maleimide groups at ends of an aliphatic, 5-atom spacer arm.
Thermo Scientific™ Pierce™ Sulfo-KMUS Crosslinker
Crosslink amines to sulfhydryls with this long crosslinker composed of sulfo-NHS-ester and maleimide groups at ends of an aliphatic, 13-atom (16.3A) spacer arm.
Thermo Scientific™ Pierce™ Sulfo-GMBS Crosslinker
Crosslink amines to sulfhydryls with this short, water-soluble crosslinker composed of sulfo-NHS-ester and maleimide groups at ends of an aliphatic, 6-atom (7.3A) spacer arm.
Thermo Scientific™ Pierce™ SMPH Crosslinker
Crosslink amines to sulfhydryls with this long crosslinker composed of NHS-ester and maleimide groups at ends of an aliphatic, 12-atom (14.2A) spacer arm.
Thermo Scientific™ Pierce™ Sulfo-MBS Crosslinker
Crosslink amines to sulfhydryls with this short, water-soluble crosslinker composed of sulfo-NHS-ester and maleimide groups at ends of an aromatic, 6-atom (7.3A) spacer arm.
Thermo Scientific™ Pierce™ Sulfo-SMPB Crosslinker
Crosslink amines to sulfhydryls with this mid-length, water-soluble crosslinker composed of sulfo-NHS-ester and maleimide groups at ends of an aromatic, 10-atom spacer arm.
Thermo Scientific™ Pierce™ EMCS Crosslinker Available on GSA/VA Contract for Federal Government customers only.
Crosslink amines to sulfhydryls with this mid-length crosslinker composed of NHS-ester and maleimide groups at ends of an aliphatic, 8-atom (9.4A) spacer arm.
Thermo Scientific™ Pierce™ Sulfo-EMCS Crosslinker
Crosslink amines to sulfhydryls with this mid-length crosslinker composed of sulfo-NHS-ester and maleimide groups at ends of a water-soluble, 8-atom (9.4A) spacer arm.
Thermo Scientific™ Pierce™ Premium Grade Sulfo-SMCC
Optimize applications where product integrity and risk minimization are paramount our highest quality formulation of this popular amine-to-sulfhydryl crosslinker.
Thermo Scientific™ Pierce™ AMAS Crosslinker
Crosslink amines to sulfhydryls with this very short crosslinker composed of NHS-ester and maleimide groups at ends of an aliphatic, 4-atom (4.4A) spacer arm.
  spinner