Sulfhydryl-to-Carbohydrate Crosslinkers
Thermo Scientific™ Pierce™ EMCH Crosslinker
Crosslink sulfhydryls to periodate-treated glycoprotein sugars with this mid-length (11.8A) crosslinker containing maleimide and hydrazide groups.
Thermo Scientific™ Pierce™ BMPH Crosslinker
Crosslink sulfhydryls to periodate-treated glycoprotein sugars with this short (8.1A) crosslinker containing maleimide and hydrazide groups.
Thermo Scientific™ Pierce™ KMUH Crosslinker
Crosslink sulfhydryls to periodate-treated glycoprotein sugars with this long (19.0A) crosslinker containing maleimide and hydrazide groups.
Thermo Scientific™ Pierce™ MPBH Crosslinker
Crosslink sulfhydryls to periodate-treated glycoprotein sugars with this long (17.9A) crosslinker containing maleimide and hydrazide groups.
Thermo Scientific™ Pierce™ PDPH Crosslinker
Crosslink sulfhydryls to periodate-treated glycoprotein sugars via cleavable disulfide bonds using this short crosslinker containing pyridyldithiol and hydrazide groups.
  spinner