Sulfhydryl-to-Sulfhydryl Crosslinkers
Thermo Scientific™ Pierce™ DTME Crosslinker
Crosslink sulfhydryls to sulfhydryls (cysteine residues) with this long, cleavable crosslinker composed of maleimide groups and 11-atom (13.3A) disulfide spacer arm.
Thermo Scientific™ Pierce™ BMOE Crosslinker
Crosslink sulfhydryls to sulfhydryls (cysteine residues in proteins or peptides) with this short crosslinker composed of maleimide groups and a 7-atom (8.0A) spacer arm.
Thermo Scientific™ Pierce™ TMEA Crosslinker
A 3-way, sulfhydryl-to-sulfhydryl crosslinker; tris (2-maleimidoethyl) amine; composed of three maleimide groups on short spacer arms; conjugate protein or peptide cysteines.
Thermo Scientific™ Pierce™ BM(PEG)3 Crosslinker
Crosslink sulfhydryls to sulfhydryls (cysteine residues) with this long, pegylated, water-soluble crosslinker composed of maleimide groups and 3-unit (17.8A) PEG spacer arm.
Thermo Scientific™ Pierce™ BMH Crosslinker
Crosslink sulfhydryls to sulfhydryls (cysteine residues in proteins or peptides) with this long crosslinker composed of maleimide groups and 11-atom (13.0A) spacer arm.
Thermo Scientific™ Pierce™ BM(PEG)2 Crosslinker
Crosslink sulfhydryls to sulfhydryls (cysteine residues) with this long, pegylated, water-soluble crosslinker composed of maleimide groups and 2-unit (14.7A) PEG spacer arm.
Thermo Scientific™ Pierce™ BMB Crosslinker
Crosslink sulfhydryls to sulfhydryls (cysteine residues in proteins or peptides) with this mid-length crosslinker composed of maleimide groups and 9-atom (10.9A) spacer arm.
Promega™ Immobilized Trypsin
For effective removal of trypsin, chymotrypsin and elastase from protein digests
  spinner