Protein Labeling Reagents
Alfa Aesar™ Bolton-Hunter Reagent
For iodination of proteins that do not contain tyrosine residues
Thermo Scientific™ EZ-Link™ Sulfo-NHS-LC-Biotin and Labeling Kits Available on GSA/VA Contract for Federal Government customers only.
Biotinylate protein amines (e.g., lysines) with this very popular mid-length (22.4A), water-soluble, NHS-ester biotinylation reagent for antibody or cell-surface labeling.
Thermo Scientific™ Pierce™ Biotin Quantitation Kit Available on GSA/VA Contract for Federal Government customers only.
HABA dye reagent and biotinylated protein standard for convenient colorimetric determination of biotinylation levels in antibodies and other proteins.
Thermo Scientific™ DyLight™ Fluor Phosphine Reagents
Label and detect azide-tagged molecules for metabolic labeling (Staudinger ligation) for fluorescence assays with phosphine-activated fluorescent dyes.
Thermo Scientific™ EZ-Link™ TFPA-PEG3-Biotin
Biotinylate molecules without common functional groups for coupling with this efficient, light-activated biotinylation reagent based on tetrafluorophenyl azide.
Thermo Scientific™ EZ-Link™ Hydrazide-LC-Biotin
Biotinylate glycoproteins (aldehydes of oxidized sugars) with this mid-length (24.7A) hydrazide-activated biotinylation reagent.
Thermo Scientific™ EZ-Link™ Sulfo-NHS-LC-Biotin No-Weigh™ Format
Thermo Scientific™ EZ-Link™ Sulfo-NHS-LC-Biotin No-Weigh™ Format is an intermediate-length, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines.
Thermo Scientific™ EZ-Link™ Biocytin
Synthesize long or trifunctional biotinylation reagents using this biotin variant containing a primary amine in its valeric acid chain.
Thermo Scientific™ EZ-Link™ Hydrazide-Biocytin
Biotinylate glycoproteins (aldehydes of oxidized sugars) with this short (19.7A) hydrazide-activated biotinylation reagent which also contains a second amine for conjugation.
Thermo Scientific™ Pierce™ Iodination Tubes
Reliably label (iodinate) proteins and peptides with iodine-125 using glass test tubes that are pre-coated (activated) with iodination reagent.
Thermo Scientific™ EZ-Link™ Amine-PEG2-Biotin
Biotinylate proteins or surfaces using EDC or other amine-reactive crosslinkers with this short (20.4A) biotinylation reagent containing a 2-unit polyethylene glycol spacer.
Thermo Scientific™ EZ-Link™ Psoralen-PEG3-Biotin
Biotinylate DNA or RNA via intercalation and conjugation with thymine- and other pyrimidine-containing bases with this light-activated biotinylation reagent.
Thermo Scientific™ EZ-Link™ NHS-PEG4-Biotin No-Weigh™ Format
Thermo Scientific™ EZ-Link™ NHS-PEG4-Biotin No-Weigh™ Format is a pegylated, water-soluble reagent for simple and efficient biotin labeling of antibodies, proteins and other primary amine-containing macromolecules.
BD 7-AAD Staining Solution
Convenient, ready-to-use nucleic acid dye that can be used in place of propidium iodide (PI)
Thermo Scientific™ EZ-Link™ Amine-PEG3-Biotin
Biotinylate proteins or surfaces using EDC or other amine-reactive crosslinkers with a mid-length (22.9A) biotinylation reagent containing a 3-unit polyethylene glycol spacer.
Thermo Scientific™ EZ-Link™ Hydrazide-PEG4-Biotin
Biotinylate glycoproteins (aldehydes of oxidized sugars) with this mid-length (31.3A), pegylated biotinylation reagent containing a 4-unit PEG spacer for solubility.
Thermo Scientific™ Pierce™ Fluorescence Biotin Quantitation Kit
Determine biotinylation levels in labeled antibodies and other proteins using this fluorescent assay reagent and HABA dye for rapid and sensitive measurement.
Thermo Scientific™ Pierce™ Iodination Beads
Easily and gently label (iodinate) proteins and peptides with iodine-125 using reagent-coated polystyrene beads that can be removed following reaction.
Thermo Scientific™ EZ-Link™ Sulfo-NHS-Biotin No-Weigh™ Format
Thermo Scientific™ EZ-Link™ Sulfo-NHS-Biotin No-Weigh™ Format is a short-chain, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines.
Thermo Scientific™ EZ-Link™ Sulfo-NHS-LC-LC-Biotin Available on GSA/VA Contract for Federal Government customers only.
Biotinylate protein amines (e.g., lysines) with this doubly long (30.5A), water-soluble, NHS-ester biotinylation reagent for antibody or cell-surface labeling.
Thermo Scientific™ EZ-Link™ Sulfo-NHS-SS-Biotin and Labeling Kits Available on GSA/VA Contract for Federal Government customers only.
Biotinylate protein amines with this cleavable, mid-length (24.3A), water-soluble, NHS-ester biotinylation reagent containing a cleavable disulfide bond in the spacer arm.
Thermo Scientific™ EZ-Link™ Maleimide-PEG2-Biotin
Biotinylate reduced cysteines with this short, maleimide-activated, sulfhydryl-reactive biotinylation reagent containing a 2-unit PEG spacer arm to enhance water solubility.
Thermo Scientific™ Pierce™ HABA Reagent
A colored, biotin assay reagent HABA (4'-hydroxyazobenzene-2-carboxylic acid) to measure biotinylation levels of biotinylated antibodies or proteins.
Thermo Scientific™ Pierce™ Iodination Reagent
Activate iodine for incorporation of iodine-125 labels into protein tyrosyl groups using this tetrachloro-diphenyl-glycoluril oxidizing agent.
Thermo Scientific™ EZ-Link™ Amine-PEG11-Biotin
Biotinylate proteins or surfaces using EDC or other amine-reactive crosslinkers with this long (53.2A) biotinylation reagent containing an 11-unit polyethylene glycol spacer.
Thermo Scientific™ EZ-Link™ Hydrazide-Biotin
Biotinylate glycoproteins (aldehydes of oxidized sugars) with this simplest and shortest (15.7A) hydrazide-activated biotinylation reagent.
Thermo Scientific™ EZ-Link™ HPDP-Biotin Available on GSA/VA Contract for Federal Government customers only.
Biotinylate reduced protein or peptide cysteines with this pyridyldithiol-activated, sulfhydryl-reactive biotinylation reagent containing a cleavable disulfide bond.
Thermo Scientific™ EZ-Link™ Alkoxyamine-PEG4-Biotin
Biotinylate glycoproteins (aldehydes of oxidized sugars) with a mid-length (27.0A), pegylated biotinylation reagent containing a 4-unit PEG spacer for length and solubility.
Thermo Scientific™ EZ-Link™ Pentylamine-Biotin
Biotinylate proteins or surfaces using EDC or other amine-reactive crosslinkers with this short (18.9A) biotinylation reagent.
Thermo Scientific Pierce™ EZ-Link™ Sulfo NHS-LC-LC-Biotin, No-Weigh Format
Thermo Scientific EZ-Link Sulfo-NHS-LC-LC-Biotin enables simple and efficient biotin labeling of antibodies, proteins, and any other primary amine–containing macromolecules.
  spinner