Amine Biotinylation Reagents and Kits
Thermo Scientific™ EZ-Link™ Sulfo-NHS-LC-Biotin and Labeling Kits Available on GSA/VA Contract for Federal Government customers only.
Biotinylate protein amines (e.g., lysines) with this very popular mid-length (22.4A), water-soluble, NHS-ester biotinylation reagent for antibody or cell-surface labeling.
Thermo Scientific™ EZ-Link™ NHS-PEG4-Biotin and Labeling Kits Available on GSA/VA Contract for Federal Government customers only.
Biotinylate protein amines and maximize solubility of proteins with this long (29.0A) NHS-ester biotinylation reagent containing a pegylated, water-soluble spacer arm.
Thermo Scientific Pierce™ EZ-Link™ Sulfo NHS-LC-LC-Biotin, No-Weigh Format
Thermo Scientific EZ-Link Sulfo-NHS-LC-LC-Biotin enables simple and efficient biotin labeling of antibodies, proteins, and any other primary amine–containing macromolecules.
Thermo Scientific™ EZ-Link™ Sulfo-NHS-SS-Biotin and Labeling Kits Available on GSA/VA Contract for Federal Government customers only.
Biotinylate protein amines with this cleavable, mid-length (24.3A), water-soluble, NHS-ester biotinylation reagent containing a cleavable disulfide bond in the spacer arm.
Thermo Scientific™ EZ-Link™ Sulfo-NHS-LC-LC-Biotin Available on GSA/VA Contract for Federal Government customers only.
Biotinylate protein amines (e.g., lysines) with this doubly long (30.5A), water-soluble, NHS-ester biotinylation reagent for antibody or cell-surface labeling.
Thermo Scientific™ EZ-Link™ Sulfo-NHS-Biotin and Labeling Kits Available on GSA/VA Contract for Federal Government customers only.
Biotinylate protein amines (e.g., lysines) with our shortest (13.5A), water-soluble, NHS-ester biotinylation reagent for antibody or cell-surface labeling.
Thermo Scientific™ EZ-Link™ NHS-Biotin
Biotinylate protein amines (e.g., lysines) with a simple, short (13.5A), NHS-ester biotinylation reagent containing a membrane-permeable spacer arm for intracellular labeling.
Thermo Scientific™ Pierce™ Antibody Biotinylation Kit for IP Available on GSA/VA Contract for Federal Government customers only.
Label primary antibodies with biotin for use in agarose or magnetic bead immunoprecipitation experiments using the Thermo Scientific™ Pierce™ Antibody Biotinylation Kit for IP.
Thermo Scientific™ EZ-Link™ NHS-LC-LC-Biotin
Biotinylate protein amines with this doubly-long-chain (30.5A), NHS-ester biotinylation reagent containing a membrane permeable spacer arm for intracellular labeling.
Thermo Scientific™ Pierce™ Premium Grade Sulfo-NHS-LC-Biotin
Optimize applications where product integrity and risk minimization are paramount with our highest quality formulation of this popular amine-reactive biotinylation reagent.
Thermo Scientific™ EZ-Link™ NHS-PEG12-Biotin
Biotinylate protein amines and maximize solubility of proteins with this very long (56.0A) NHS-ester biotinylation reagent containing a pegylated, water-soluble spacer arm.
Thermo Scientific Pierce™ EZ-Link™ NHS-PEG12-Biotin, No-Weigh Format
Thermo Scientific™ EZ-Link NHS-PEG12-Biotin enables simple and efficient biotin labeling of antibodies, proteins, and any other primary amine–containing macromolecule.
Thermo Scientific™ EZ-Link™ NHS-SS-PEG4-Biotin
Biotinylate protein amines with this cleavable, long (37.9A) NHS-ester biotinylation reagent containing a reducible disulfide bond in the pegylated, water-soluble spacer arm.
Thermo Scientific™ EZ-Link™ NHS-PEG Solid-Phase Biotinylation Kit
Biotinylate protein amines with greater control over the amine-labeling reaction and antibody cleanup process using this column-based method of antibody biotinylation.
Thermo Scientific™ EZ-Link™ NHS-PEG Solid-Phase Biotinylation Kit
A column-based approach to antibody biotinylation and clean-up
Thermo Scientific™ Pierce™ Premium Grade Sulfo-NHS-SS-Biotin
Biotin-label antibodies or other proteins and biomolecules in applications where product integrity and risk minimization are paramount with Thermo Scientific™ Pierce™ Premium Grade Sulfo-NHS-SS-Biotin, a high-quality formulation of this amine-reactive, cleavable biotinylation reagent.
Thermo Scientific™ EZ-Link™ TFP-PEG3-Biotin
Biotinylate primary amines (e.g., protein lysines) and secondary amines such as histidine and tryptophan with this tetrafluorophenyl-ester biotinylation reagent.
Thermo Scientific™ EZ-Link™ NHS-LC-Biotin
Biotinylate protein amines (e.g., lysines) with this long-chain (22.4A), NHS-ester biotinylation reagent containing a membrane permeable spacer arm for intracellular labeling.
Thermo Scientific™ EZ-Link™ PFP-Biotin
Biotinylate primary amines in proteins as well as secondary amines in DNA or RNA with this very short (9.6A) pentafluorophenyl-ester biotinylation reagent.
Thermo Scientific™ EZ-Link™ NHS-SS-Biotin
Biotinylate protein amines with this cleavable, mid-length (24.3A), NHS-ester biotinylation reagent containing a reducible disulfide bond in the membrane-permeable spacer arm.
Abnova™ Biotin Labeling Kit Available on GSA/VA Contract for Federal Government customers only.
Primarily used for the preparation of biotin-labeled antibody for enzyme immunoassay (EIA)
  spinner