Biotin Quantitation Reagents and Kits
Thermo Scientific™ Pierce™ Biotin Quantitation Kit Available on GSA/VA Contract for Federal Government customers only.
HABA dye reagent and biotinylated protein standard for convenient colorimetric determination of biotinylation levels in antibodies and other proteins.
Thermo Scientific™ Pierce™ Fluorescence Biotin Quantitation Kit
Determine biotinylation levels in labeled antibodies and other proteins using this fluorescent assay reagent and HABA dye for rapid and sensitive measurement.
Thermo Scientific™ Pierce™ HABA Reagent
A colored, biotin assay reagent HABA (4'-hydroxyazobenzene-2-carboxylic acid) to measure biotinylation levels of biotinylated antibodies or proteins.
Molecular Probes™ FluoReporter™ Biotin Quantitation Assay Kit, for biotinylated proteins Non-distribution product as customer accommodation.
FluoReporter™ Biotin Quantitation Assay Kit, for biotinylated proteins
  spinner