Carbohydrate Biotinylation Reagents
Thermo Scientific™ EZ-Link™ Hydrazide-Biocytin
Biotinylate glycoproteins (aldehydes of oxidized sugars) with this short (19.7A) hydrazide-activated biotinylation reagent which also contains a second amine for conjugation.
Thermo Scientific™ EZ-Link™ Hydrazide-LC-Biotin
Biotinylate glycoproteins (aldehydes of oxidized sugars) with this mid-length (24.7A) hydrazide-activated biotinylation reagent.
Thermo Scientific™ EZ-Link™ Hydrazide-Biotin
Biotinylate glycoproteins (aldehydes of oxidized sugars) with this simplest and shortest (15.7A) hydrazide-activated biotinylation reagent.
Thermo Scientific™ EZ-Link™ Hydrazide-PEG4-Biotin
Biotinylate glycoproteins (aldehydes of oxidized sugars) with this mid-length (31.3A), pegylated biotinylation reagent containing a 4-unit PEG spacer for solubility.
Thermo Scientific™ EZ-Link™ Alkoxyamine-PEG12-Biotin
Biotinylate glycoproteins (aldehydes of oxidized sugars) with a very long (55.4A), pegylated biotinylation reagent containing a 12-unit PEG spacer for length and solubility.
Thermo Scientific™ EZ-Link™ Alkoxyamine-PEG4-Biotin
Biotinylate glycoproteins (aldehydes of oxidized sugars) with a mid-length (27.0A), pegylated biotinylation reagent containing a 4-unit PEG spacer for length and solubility.
  spinner