Carboxyl Biotinylation Reagents
Thermo Scientific™ EZ-Link™ Amine-PEG3-Biotin
Biotinylate proteins or surfaces using EDC or other amine-reactive crosslinkers with a mid-length (22.9A) biotinylation reagent containing a 3-unit polyethylene glycol spacer.
Thermo Scientific™ EZ-Link™ Amine-PEG2-Biotin
Biotinylate proteins or surfaces using EDC or other amine-reactive crosslinkers with this short (20.4A) biotinylation reagent containing a 2-unit polyethylene glycol spacer.
Thermo Scientific™ EZ-Link™ Biocytin
Synthesize long or trifunctional biotinylation reagents using this biotin variant containing a primary amine in its valeric acid chain.
Thermo Scientific™ EZ-Link™ Amine-PEG11-Biotin
Biotinylate proteins or surfaces using EDC or other amine-reactive crosslinkers with this long (53.2A) biotinylation reagent containing an 11-unit polyethylene glycol spacer.
Thermo Scientific™ EZ-Link™ Pentylamine-Biotin
Biotinylate proteins or surfaces using EDC or other amine-reactive crosslinkers with this short (18.9A) biotinylation reagent.
Thermo Scientific™ EZ-Link™ Hydrazide-PEG4-Desthiobiotin No-Weigh™ Format
Thermo Scientific™ EZ-Link™ Hydrazide-PEG4-Desthiobiotin No-Weigh™ Format is a pegylated, carbonyl-reactive labeling reagent whose biotin-like group is elutable from streptavidin, enabling its use for cell surface glycoprotein labeling and purification.
  spinner