Metabolic Labeling Reagents for Proteins
Thermo Scientific™ DyLight™ Fluor Phosphine Reagents
Label and detect azide-tagged molecules for metabolic labeling (Staudinger ligation) for fluorescence assays with phosphine-activated fluorescent dyes.
Thermo Scientific™ Pierce™ Azido-Sugars
Study glycoproteins using in vivo metabolic labeling and chemoselective ligation to phosphine reagents with these azide-tagged sugars (GlcNAz, GalNAz, ManNAz).
Thermo Scientific™ Pierce™ Biotin-PEG3-Phosphine
Label azide-tagged molecules for metabolic labeling (Staudinger ligation) and streptavidin detection using this phosphine-activated biotinylation reagent.
MilliporeSigma™ Calbiochem™ β-Cyclodextrin
Homogeneous cyclic α1,4-linked D-glucopyranose units in a seven-member ring. Forms clathrates. Suitable for use with dansyl chloride to form water-soluble complexes for fluorescent labeling of proteins.
  spinner