Pegylation Reagents
Thermo Scientific™ Pierce™ Methyl-PEG-NHS-Ester Reagents
Pegylate amines (-NH2 groups) on proteins (lysine residues) or assay surfaces using MS(PEG)n, amine-reactive (NHS ester), methyl-capped PEG compounds (4 to 24 EG units).
Thermo Scientific™ Pierce™ Carboxy-PEG-Thiol Compounds
CT(PEG)n: carboxylic acid and sulfhydryl compound with a 12-unit polyethylene glycol (PEG) spacer; carboxylate and pegylate quantum dots, gold surfaces, silver.
Thermo Scientific™ Pierce™ Methyl-PEG-Amine Compounds
MA(PEG)n: methyl- and amine-capped polyethylene glycol compounds containing 4 to 24 PEG units; use for surface-modification and molecule-pegylation applications.
Thermo Scientific™ Pierce™ Carboxy-PEG-Amine Compounds
CA(PEG)n: pegylated amino acids of the form carboxy-PEG-amine; with 4 to 24 polyethylene glycol units; for surface- and molecule-pegylation applications.
Thermo Scientific™ Pierce™ Methyl-PEG-Lipoamide Compounds
ML(PEG)n: methyl and bidentate thiol compound with a 4-unit polyethylene glycol (PEG) spacer; methylate and pegylate quantum dots, gold surfaces, silver.
Thermo Scientific™ Pierce™ Carboxy-PEG-Lipoamide Compounds
CL(PEG)n: carboxylic acid and bidentate thiol compound with a 4-unit polyethylene glycol (PEG) spacer; carboxylate and pegylate quantum dots, gold surfaces, silver.
Thermo Scientific™ Pierce™ Methyl-PEG-Maleimide Reagents
Pegylate sulfhydryls (-SH groups) on proteins (cysteine residues) or assay surfaces using MS(PEG); sulfhydryl-reactive (maleimide), PEG compounds (12 and 24 EG units).
Thermo Scientific™ Pierce™ Methyl-PEG-Thiol Compounds
MT(PEG)n: methyl and sulfhydryl compound with a 4-unit polyethylene glycol (PEG) spacer; methylate and pegylate quantum dots, gold surfaces, silver.
  spinner