Proteases and Protein-Cleaving Reagents
Thermo Scientific™ Pierce™ Proteases for Mass Spectrometry Available on GSA/VA Contract for Federal Government customers only.
Improve sequence coverage and protein identification for mass spectrometry using these individual, highly specific proteases in custom combinations.
Thermo Scientific™ Pierce™ Proteinase K Available on GSA/VA Contract for Federal Government customers only.
Cleave at the carboxyl-side of aliphatic, aromatic or hydrophobic residues and digest-inactivate DNase and RNase in nucleic acid purification using this protease.
Thermo Scientific™ Pierce™ Human Rinovirus 3C Protease (HRV 3C) Available on GSA/VA Contract for Federal Government customers only.
Remove fusion tags from proteins with the HRV 3C cleavage sequence with this recombinant cysteine protease that is dual tagged for easy removal from the sample after cleavage.
Thermo Scientific™ DSSO (disuccinimidyl sulfoxide)
MS-cleavable and membrane-permeable crosslinker that contains amine-reactive N-hydroxysuccinimide (NHS) ester at each end of a 7-carbon spacer arm.
Thermo Scientific™ Pierce™ Immobilized Papain Available on GSA/VA Contract for Federal Government customers only.
Generate and purify Fab fragments from antibodies with cysteine-endopeptidase papain immobilized onto beaded agarose resin.
Thermo Scientific™ TMTsixplex™ Isobaric Label Reagent Set
TMT™ Isobaric Mass Tagging Kits and Reagents enable multiplex quantitation of proteins extracted from cells and tissues using tandem mass spectrometry.
Thermo Scientific™ Pierce™ Immobilized Pepsin Available on GSA/VA Contract for Federal Government customers only.
Generate and purify Fab and F(ab')2 fragments from antibodies with acidic endopeptidase (pepsin) immobilized onto beaded agarose resin.
Thermo Scientific™ Pierce™ In-Gel Tryptic Digestion Kit Available on GSA/VA Contract for Federal Government customers only.
Trypsin-digest and recover peptide fragments for mass spectrometry from 150 stained protein bands excised from SDS-PAGE (polyacrylamide gel plugs).
Promega™ IdeS Protease
Antibody fragmentation and characterization, simplified.
EMD Millipore™ HRV 3C Protease Available on GSA/VA Contract for Federal Government customers only.
Highly efficient fusion tag removal at low temperature
Thermo Scientific™ Pierce™ TPCK Trypsin and Resin
Cleave the carboxyl-side of Arg and Lys residues with trypsin that isTPCK-treated to block chymotrypsin activity; its immobilized form allows enzyme removal after digestion.
MP Biomedicals™ Pepsin
From porcine stomach mucosa
Trypsin, porcine pancreas
A pancreatic serine protease
Promega Caspase-Glo 1 Inflammasome Assay, 10ml
The Caspase-Glo 1 Inflammasome Assay is a homogeneous, bioluminescent method to selectively measure the activity of caspase-1, a member of the cysteine aspartic acid-specific protease (caspase) family and an essential component of the inflammasome.
Thermo Scientific™ Pierce™ Immobilized Ficin
Enable controlled antibody fragmentation, especially of mouse IgG1, with this sulfhydryl-specific ficin protease immobilized onto beaded agarose resin.
Promega IdeS Protease, Frozen, 2,000 units
IdeS Protease is an immunoglobulin-degrading enzyme from Streptococcus pyogenes (IdeS). IdeZ Protease is an immunoglobulin-degrading enzyme from Streptococcus equi subspecies zooepidemicus.
Promega AccuMAP Low pH Protein Digestion Mini Kit, 1 each
AccuMAP Low pH Protein Digestion Kit enables suppression of sample preparation-induced artificial deamidation, disulfide bond scrambling and oxidation overthe course of sample preparation. Pre-existing non-enzymatic modifications remain intact.
EMD Millipore™ Thrombin, Restriction Grade Available on GSA/VA Contract for Federal Government customers only.
Prepared from human plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV
Thermo Scientific™ Pierce™ In-Solution Tryptic Digestion and Guanidination Kit
Alkylate and digest proteins and then guanidinate the peptide fragments for mass spectrometry (MS) using proteomics-grade trypsin, buffers and reagents.
Affymetrix/USB™ Pepsin from Porcine Stomach
Derived from Porcine stomach
EMD Millipore™ Proteinase K Solution, 600mAU/mL Available on GSA/VA Contract for Federal Government customers only.
Efficient removal of proteins from nucleic acid solutions
Pepstatin A, 98%
Protease inhibitor that acts on pepsin, renin and cathepsin D
Promega™ ProTEV Plus Protease
Engineered protease with greater stability and prolonged activity over native TEV protease.
EMD Millipore™ Endoproteinase Asp-N, Excision Grade, Pseudomonas fragi Available on GSA/VA Contract for Federal Government customers only.
Designed for protein sequencing or sequence verification, analysis of protein structural domains, and cleavage of fusion proteins
EMD Millipore™ Calbiochem™ Carboxypeptidase B, Porcine Pancreas
Native carboxypeptidase B catalyzes the release of C-terminal from basic amino acids, arginine, lysine, or ornithine from polypeptides
Thermo Scientific™ Pierce™ Staphylococcus aureus V-8 Protease and Kt Available on GSA/VA Contract for Federal Government customers only.
Specifically cleave only at glutamic acid residues or cleave at both glutamic and aspartic residues with our immobilized Staphylococcus aureus V-8 protease kit.
EMD Millipore™ Proteinase K, Lyophilized Available on GSA/VA Contract for Federal Government customers only.
Highly active serine protease that exhibits broad cleavage specificity on native and denatured proteins and is widely used in the purification of DNA and RNA
MP Biomedicals™ Trypsin-EDTA Solution
widely used for cell dissociation, routine cell culture passaging, and primary tissue dissociation due to its digestive strength.
Promega™ Glycosidases
Characterize protein glycosylation.
  spinner