Protein Denaturants and Chaotropes
Thermo Scientific™ Pierce™ Urea
Pure, crystalline urea (CH4N2O), CAS 57-13-6; denaturant with low UV-absorptivity for protein biology methods (elution, solubilization, refolding, storage).
Thermo Scientific™ Pierce™ 8M Guanidine·HCl Solution
Ready-to-use, 8M solution of guanidinium hydrochloride (GdnCl, GuHCl), CAS 50-01-1; dilute and pH-adjust to any concentration for protein biology methods.
Thermo Scientific™ Pierce™ Guanidine·HCl
Pure, crystalline guanidium hydrochloride (GdnHCl, GuHCl), CAS 50-01-1; denaturant for protein biology methods (elution, solubilization, refolding, storage).
  spinner