Reducing Agents for Protein Disulfides
Thermo Scientific™ Bond-Breaker™ Neutral pH TCEP Solution Available on GSA/VA Contract for Federal Government customers only.
Efficiently reduce protein disulfide bonds with this stable, odorless, 0.5M solution of TCEP reducing agent that is ideal for making reducing SDS-PAGE sample buffers.
Thermo Scientific™ Pierce™ DTT (Dithiothreitol) Available on GSA/VA Contract for Federal Government customers only.
Reduce disulfides in biological buffers and sample loading buffers with pure, crystalline dithiothreitol (DTT), the most popular protein disulfide reducing agent.
Thermo Scientific™ Pierce™ TCEP·HCl Available on GSA/VA Contract for Federal Government customers only.
Reduce protein and peptide disulfide bonds using this thiol-free, pure, crystalline tris (2-carboxyethyl) phosphine hydrochloride, TCEP (CAS 5961-85-3).
Thermo Scientific™ Pierce™ 2-Mercaptoethylamine·HCl
Pure, crystalline 2-aminoethanethiol (2-MEA-HCl, also called cysteamine-HCl), CAS 156-57-0; selectively reduces antibody hinge-region disulfide bonds.
Thermo Scientific™ Pierce™ Immobilized TCEP Disulfide Reducing Gel Available on GSA/VA Contract for Federal Government customers only.
Efficiently reduce protein or peptide disulfide bonds with this stable, odorless, beaded agarose resin on which Tris (2-carboxyethyl) phosphine is immobilized.
Thermo Scientific™ Pierce™ Premium Grade TCEP-HCl
Optimize applications where product integrity and risk minimization are paramount with our highest quality formulation of this disulfide reducing agent.
Thermo Scientific™ Pierce™ 2-Mercaptoethanol
Pure, liquid (equals 14M), beta-mercaptoethanol (bME, 2BME, 2-ME, b-mer), CAS 60-24-2; thiol reducing agent for cleaving protein disulfide bonds (cystines cysteines).
Thermo Scientific™ Pierce™ Cysteine·HCl
Make sulfhydryl assay standards or perform protein refolding experiments using this pure, crystalline cysteine hydrochloride (Cys-HCl) CAS 52-89-1.
Thermo Scientific Pierce™ TCEP-HCl, No-Weigh Format
Thermo Scientific™ Pierce™ TCEP-HCl is potent, versatile, odorless, thiol-free reducing agent with broad application to protein and other research involving reduction of disulfide bonds.
Thermo Scientific™ Pierce™ Immobilized Reductant Columns
Efficiently reduce protein or peptide disulfide bonds with this reusable column on with an immobilized thiol-based reducing agent for solid-phase disulfide bond reduction.
EMD Millipore™ Calbiochem™ TCEP, Hydrochloride
Reduces even the most stable water-soluble alkyl disulfides at pH 4.5 within 5 minutes
EMD Millipore™ Calbiochem EGTA/AM Available on GSA/VA Contract for Federal Government customers only.
Application in Life Science research
EMD Millipore™ Calbiochem TCEP, Neutral Available on GSA/VA Contract for Federal Government customers only.
Ready-to-use stable, neutral form of popular disulfide-reducing agent TCEP, Hydrochloride
EMD Millipore™ Calbiochem™ PIH Available on GSA/VA Contract for Federal Government customers only.
Cell-permeable, non-toxic tridendate iron (Fe3+) chelator of aroyl hydrazone class
EMD Millipore™ Calbiochem™ TPEN Available on GSA/VA Contract for Federal Government customers only.
Lipid-soluble heavy metal chelator that crosses artificial and natural membranes
EMD Millipore™ Calbiochem™ SPUTOLYSIN™ Reagent Available on GSA/VA Contract for Federal Government customers only.
Intended for isolation of epithelial cells, pathogenic or saprophytic bacteria, fungi, and yeasts from sputum
EMD Millipore™ Calbiochem™ BAPTA/AM Available on GSA/VA Contract for Federal Government customers only.
Can be loaded into wide variety of cells, where it is hydrolyzed by cytosolic esterases and is trapped intracellularly as the active chelator BAPTA
EMD Millipore™ Calbiochem™ DM-NITROPHEN™ Reagent, Tetrasodium Salt
Caged Ca2+ chelator that undergoes a major and rapid decrease in Ca2+-binding affinity upon photolysis
  spinner