General Dialysis Supplies
Thermo Scientific™ SnakeSkin™ Dialysis Tubing Clips Available on GSA/VA Contract for Federal Government customers only.
Securely close dialysis tubing up to 50mm flat diameter, including SnakeSkin Dialysis Tubing, with these sturdy, non-floating tube closures.
Spectrum™ Spectrum™ RC Dialysis Membranes 12,000 to 14,000 Dalton MWCO
Multipurpose dialysis tubing is ideal for general dialysis for a variety of applications and volumes
Spectrum™ Spectra/Por™ Float-A-Lyzer™ G2 Dialysis Devices
Ready-to-use and disposable, Float-A-Lyzer G2 incorporates biotech-grade cellulose ester (CE) membrane tubing into a preasembled dialysis device that is easy-to-use and provides ultimate convenience for small volume dialysis (1 to 10mL)
Spectrum™ Spectrum™ Dialysis Tubing Closures: Spectra/Por™ Standard Type
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
GE Healthcare Mini Dialysis Kits Available on GSA/VA Contract for Federal Government customers only.
Offer a simple solution to the handling problems of small volume dialysis. The dialysis tubes in the kit each have a cap that incorporates a small disk of dialysis membrane.
Spectrum™ Spectra/Por™ Weighted Paired Closures
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
Spectrum™ Micro Float-A-Lyzer™ Ready-to-Use Small Volume Dialysis Device
Providing ultimate convenience, this preassembled device incorporates Spectrum's ultrapure Biotech Grade Cellulose Ester (CE) membrane tubing into an easy-to-use design making laboroatory microdialysis easier than ever
Spectrum™ Spectra/Por™ Weighted Closures
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
Spectrum™ Universal Dialysis Tubing Closures (for all Membrane Types)
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
Spectrum™ Spectra/Por™ Magnetic Weighted Closures
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
Bel-Art™ SP Scienceware™ Equilibrium Dialysis Cells Available on GSA/VA Contract for Federal Government customers only.
Feature stainless steel screws that tighten down on the dialysis membrane also acting as a gasket
Spectrum™ Dialysis Reservoir
Provides the optimum environment for laboratory dialysis
Spectrum™ Spectra/Por™ 1-4 Standard RC Dialysis Membrane in Flat Sheets
Clear, flexible and sturdy Standard RC (regenerated cellulose) in single layer square sheets that can be used with a variety of dialysis equipment or can be cut into specific dimensions for attaching to support frames for customized diagnostic assays, equilibrium studies, desalting, protein purification, etc.
Spectrum™ Spectra/Por™ Dialysis Tubing Closures for Standard RC Membrane Only
Floating closures that aid in buoyancy and vertical orientation when applied to top end of tubing
  spinner