Ion Exchange Columns
Thermo Scientific™ Pierce™ Strong Ion Exchange Spin Columns
Perform rapid protein fractionation and sample preparation based on charge differences using these strong cation and anion exchange spin columns.
GE Healthcare HiTrap™ IEX HP Prepacked Columns Available on GSA/VA Contract for Federal Government customers only.
Use for high-resolution, small-scale protein purification. GE Healthcare Life Sciences™ HiTrap™ IEX HP Prepacked Columns are prepacked, ready-to-use Sepharose™ High Performance strong cation exchange columns.
GE Healthcare HiTrap™ Q Sepharose Fast Flow IEX Columns Available on GSA/VA Contract for Federal Government customers only.
Use for small-scale protein purifications.   GE Healthcare Life Sciences™ HiTrap™ Q Sepharose Fast Flow IEX Columns are strong anion exchangers for rapid separations either alone or connected in series.
GE Healthcare HiTrap™ SP Sepharose Fast Flow IEX Columns Available on GSA/VA Contract for Federal Government customers only.
Use for small-scale protein purifications GE Healthcare Life Sciences™ HiTrap™ SP Sepharose Fast Flow IEX Columns are strong cation exchanger columns for rapid separations either alone or connected in series.
GE Healthcare HiPrep™ Q Fast Flow 16/10 Column Available on GSA/VA Contract for Federal Government customers only.
Provide fast, preparative separations of proteins and other biomolecules
GE Healthcare HiTrap™ DEAE Sepharose Fast Flow IEX Columns Available on GSA/VA Contract for Federal Government customers only.
Weak anion exchanger for small-scale protein purifications.  GE Healthcare Lifescience™ HiTrap™ DEAE Sepharose Fast Flow IEX Columns are convenient and rapid.
GE Healthcare HisPrep Column Available on GSA/VA Contract for Federal Government customers only.
Optimize purification of histidine-tagged recombinant proteins.   GE Healthcare Life Sciences™ HisPrep Column is a ready to use column offering convenient scale-up purification of histidine-tagged proteins.
GE Healthcare HiPrep™ Q HP 16/10 Column Available on GSA/VA Contract for Federal Government customers only.
Provide fast, preparative separations of proteins and other biomolecules
GE Healthcare HiPrep™ Heparin Fast Flow 16/10 Column Available on GSA/VA Contract for Federal Government customers only.
For preparative purification of proteins with affinity for heparin
GE Healthcare RESOURCE™ Q IEX Columns Available on GSA/VA Contract for Federal Government customers only.
Use for intermediate lab-scale purification and large-scale polishing of biomolecules GE Healthcare Life Sciences™ RESOURCE™ Q IEX Columns are strong anion exchanger columns.
GE Healthcare HiPrep™ Q XL 16/10 Column Available on GSA/VA Contract for Federal Government customers only.
Provide fast, preparative separations of proteins and other biomolecules
GE Healthcare HiPrep™ SP XL 16/10 Column Available on GSA/VA Contract for Federal Government customers only.
Provide fast, preparative separations of proteins and other biomolecules
GE Healthcare HiTrap™ Capto™ Q ImpRes IEX Columns Available on GSA/VA Contract for Federal Government customers only.
High-throughput medium for intermediate and polishing protein purification
GE Healthcare HiTrap™ Capto™ S IEX Columns Available on GSA/VA Contract for Federal Government customers only.
Strong sulfoethyl(S) cation exchanger
GE Healthcare HiTrap™ IEX Selection Kit Available on GSA/VA Contract for Federal Government customers only.
Use for fast and easy screening of a variety of different media. HiTrap™ IEX Selection Kits contain Sepharose™ Fast Flow and Sepharose XL in combination with seven different ion exchange ligands.
GE Healthcare RESOURCE™ S IEX Columns Available on GSA/VA Contract for Federal Government customers only.
RESOURCE prepacked columns for fast separation. Resource S IEX Columns are prepacked with Source 15S, a strong cation exchanger for intermediate lab-scale purification and large-scale polishing of biomolecules with ion exchange chromatography.
GE Healthcare HiPrep™ SP HP 16/10 Column Available on GSA/VA Contract for Federal Government customers only.
Provide fast, preparative separations of proteins and other biomolecules
GE Healthcare HiTrap™ Capto™ SP ImpRes IEX Columns Available on GSA/VA Contract for Federal Government customers only.
High-throughput medium for intermediate and polishing protein purification
GE Healthcare HiPrep™ SP Fast Flow 16/10 Column Available on GSA/VA Contract for Federal Government customers only.
Provide fast, preparative separations of proteins and other biomolecules
GE Healthcare HiTrap™ Q Sepharose XL IEX Columns Available on GSA/VA Contract for Federal Government customers only.
Optimized for fast, convenient small-scale protein capture
GE Healthcare HiTrap™ Capto™ Q IEX Columns Available on GSA/VA Contract for Federal Government customers only.
Strong quaternary ammonium (Q) anion exchanger
GE Healthcare HiTrap™ ANX Sepharose Fast Flow IEX Columnns Available on GSA/VA Contract for Federal Government customers only.
Weak anion exchanger for small scale purification of higher molecular weight protein
GE Healthcare HiTrap™ CM Sepharose Fast Flow IEX Columns Available on GSA/VA Contract for Federal Government customers only.
Weak cation exchanger for small-scale protein purifications
GE Healthcare HiTrap™ SP Sepharose XL IEX Columns Available on GSA/VA Contract for Federal Government customers only.
Optimized for fast, convenient small-scale protein capture
GE Healthcare HiTrap Capto DEAE Available on GSA/VA Contract for Federal Government customers only.
Reduce process cycle times and improve productivity for screening and small-scale protein purification. GE Healthcare HiTrap Capto DEAE combines the properties of high binding capacity, high flow rate and low back pressure. It is an excellent choice for process-scale purification.
GE Healthcare Mono Q HR 16/10 Available on GSA/VA Contract for Federal Government customers only.
Offers high capacity during the polishing step in purification of biomolecules. GE Healthcare Mono Q HR 16/10 is a strong anion exchanger packed with MonoBeads in high-volume beds. It is an excellent choice when high purity is needed.
GE Healthcare Capto Q ImpRes, 25mL Available on GSA/VA Contract for Federal Government customers only.
Designed to meet the demands of modern, large-scale manufacturers. GE Healthcare Capto Q ImpRes, 25ml is a strong anion exchange BioProcess medium that enables fast, efficient, and cost-effective intermediate and polishing protein purification.
GE Healthcare Mini Q PC 3.2/3 Available on GSA/VA Contract for Federal Government customers only.
Improves resolution, withstands back pressure and minimizes nonspecific adsorption. GE Healthcare Mini Q PC 3.2/3 precision columns are prepacked with MiniBeads for microscale final polishing and analytical ion exchange separations.
GE Healthcare Capto DEAE, 25mL Available on GSA/VA Contract for Federal Government customers only.
Designed to meet the demands of modern large-scale manufacturers. GE Healthcare Capto DEAE, 25ml is a weak anion exchange BioProcess medium for fast, efficient, and cost-effective capture and intermediate protein purification.
  spinner