Coated Microplates for ELISA
Thermo Scientific™ Pierce™ High Binding Streptavidin Coated Plates Available on GSA/VA Contract for Federal Government customers only.
Bind biotinylated oligonucleotides and peptides with higher binding efficiency than other commercially available streptavidin-coated plates.
Thermo Scientific™ Pierce™ Nickel Coated Plates Available on GSA/VA Contract for Federal Government customers only.
Bind His-tagged fusion proteins for ELISA and other plate-based assays to study interactions of expressed recombinant proteins.
Thermo Scientific™ Pierce™ High Binding NeutrAvidin™ Coated Plates
Attain higher biotin-binding efficiency with these pre-blocked, ready-to-use high-capacity NeutrAvidin protein-coated microplates.
Thermo Scientific™ Pierce™ Standard Binding Streptavidin Coated Plates
Capture biotinylated antibodies or probes for ELISA and other target-specific assays with pre-blocked streptavidin-coated plates.
Thermo Scientific™ Pierce™ Protein A, G, and L Coated Plates
Capture antibody complexes with these alternatives to direct, passive adsorption for immobilizing antibodies for ELISA and other plate-based assays.
Thermo Scientific™ Pierce™ Standard Binding NeutrAvidin Coated Plates
Capture biotinylated antibodies or probes for ELISA and other target-specific assays with pre-blocked NeutrAvidin protein-coated plates.
Thermo Scientific™ Pierce™ Maleimide Activated (Sulfhydryl-binding) Plates
Covalently attach sulfhydryl-containing molecules that are difficult to coat onto polystyrene plates, such as peptides that contain a terminal cysteine.
Thermo Scientific™ Pierce™ Glutathione Coated Plates
Capture and detect glutathione-S-transferase (GST) fusion proteins with these pre-blocked and glutathione (GSH)-coated microplates.
Thermo Scientific™ Pierce™ Maleic Anhydride (Amine-binding) Plates
Bind amine-containing peptides and other molecules to microplate wells for use in binding assays with these maleic anhydride-activated plates.
Thermo Scientific™ Pierce™ Biotin Coated Plates
Capture NeutrAvidin protein, streptavidin, avidin or any other biotin-binding protein with these biotin-coated plates.
Thermo Scientific™ Pierce™ High Capacity Copper Coated Plates
Analyze polyhistidine-tagged fusion proteins by ELISA with these high-capacity plates treated to maximize histidine-tagged protein binding.
Thermo Scientific™ Pierce™ Anti-GST Coated Plates
Detect glutathione S-transferase (GST)-tagged proteins with these ready-to-use pre-blocked, mouse monoclonal anti-GST antibody coated polystyrene microplates.
Thermo Scientific™ Pierce™ Secondary Antibody Coated Plates
Perform binding assays when available antibodies are in low quantities or they denature and become inactive upon direct adsorption to polystyrene plates.
Thermo Scientific™ Pierce™ High Sensitivity NeutrAvidin Coated Plates
Maximize biotinylated protein capture efficiency and control background with microplates coated to detect low concentrations of biotinylated analytes.
Thermo Scientific™ Pierce™ High Sensitivity Streptavidin Coated Plates
Maximize biotinylated protein capture efficiency and control background with microplates coated to detect low concentrations of biotinylated analytes.
Thermo Scientific™ Pierce™ Biotin-Binding Plate Sample Pack
Determine which variety of streptavidin-coated plate is best for a particular assay involving biotinylated molecules with this sample pack.
Thermo Scientific™ Pierce™ Maleic Anhydride (Amine-binding) Plates
Covalently bind proteins, peptides and other amine-containing compounds
  spinner