GM-CSF ELISA Kits
R&D Systems™ Human GM-CSF Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
A 3.5 or 4.5 hour solid phase ELISA designed to measure GM-CSF levels in cell culture supernates, serum, and plasma. It contains E. coli-expressed recombinant human GM-CSF and antibodies raised against the recombinant factor.
R&D Systems™ Mouse GM-CSF Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid phase ELISA designed to measure mouse GM-CSF in cell culture supernates and serum.
R&D Systems™ Mouse GM-CSF DuoSet ELISA Kit
Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
Invitrogen™ Rat CSF2 (GM-CSF) ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay rat CSF2 (GM-CSF) levels in cell culture supernatants, plasma, serum samples.
Invitrogen™ Swine CSF2 (GM-CSF) ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay swine CSF2 (GM-CSF) levels in cell culture supernatants, plasma, serum samples.
R&D Systems™ Human GM-CSF DuoSet ELISA Kit
Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
R&D Systems™ Human GM-CSF Quantikine HS ELISA Kit, R&D Systems™
a 6.5 hour solid phase ELISA designed to measure human GM-CSF in serum, plasma, and urine.
Invitrogen™ Human GM-CSF ELISA Kits Non-distribution product as customer accommodation.
Granulocyte Macrophage Colony Stimulating Factor Recombinant protein SGMCSF detects Granulocyte Macrophage Colony Stimulating Factor in Not Applicable samples, and is validated for Control, ELISA and WB
Invitrogen™ Mouse GM-CSF ELISA Kits Non-distribution product as customer accommodation.
Measure levels of granulocyte macrophage colony-stimulating factor in mouse serum and culture supernatants using 96-well plates.
  spinner