IL-12 ELISA Kits
Human IL-12 p70 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 3.5 or 4.5 hour solid phase ELISA designed to measure IL-12 in cell culture supernates, serum, plasma, and urine.
Mouse IL-12 p70 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid phase ELISA designed to measure mouse IL-12 p70 in cell culture supernates and mouse serum.
eBioscience™ Human IL-12 p70 ELISA Ready-SET-Go!™ Kit Non-distribution product as customer accommodation.
Specifically measures the bioactive, heterodimeric form of IL-12, p70, without interference by p40 monomer, homodimer, or IL-23 (p19/p40). Engineered for accurate and precise measurement of human IL-12 p70 protein levels from samples including serum, plasma, and cell culture supernatants.
eBioscience™ Mouse IL-12/IL-23 total p40 ELISA Ready-SET-Go!™ Kit Non-distribution product as customer accommodation.
Specifically engineered for accurate and precise measurement of mouse IL-12 / IL-23 (total p40) protein levels from samples including serum, and cell culture supernatants.
eBioscience™ Mouse IL-12 p70 ELISA Ready-SET-Go!™ Kit Non-distribution product as customer accommodation.
Specifically measures the bioactive, heterodimeric form of IL-12, p70, without interference by p40 monomer or the related protein IL-23 (p19/p40). Engineered for accurate and precise measurement of mouse IL-12 p70 protein levels from samples including serum, and cell culture supernatants.
Invitrogen™ Novex EASIA IL-12 p40 Human ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Designed to quantify protein levels of human IL-12 p35/p40 heterodimer (or IL-12p70)
Invitrogen™ Mouse IL-12 (p40) ELISA Kits Non-distribution product as customer accommodation.
Measure levels of inactive mouse interleukin-12, p40 in mouse serum, plasma and culture supernatant samples using 96-well plates.
eBioscience™ Human IL-12 (p70) Platinum ELISA Kit Non-distribution product as customer accommodation.
For the quantitative detection of human IL-12 p70.
Invitrogen™ Mouse IL-12 (p70) ELISA Kits Non-distribution product as customer accommodation.
Measure levels of active mouse interleukin-12, p70 in mouse serum and culture supernatant samples using 96-well plates.
Invitrogen™ Novex IL-12 p70 Human ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Designed to quantify protein levels
Invitrogen™ Novex IL-12 Human ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Quantifies natural and recombinant human IL-12 p35/p40 heterodimer (or IL-12p70)
eBioscience™ Mouse IL-12 p70 Platinum ELISA Kit Non-distribution product as customer accommodation.
For the quantitative detection of mouse IL-12 p70.
Novus Biologicals™ Mouse IL-23 ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
eBioscience™ Human IL-12/IL-23 p40 Platinum ELISA Kit Non-distribution product as customer accommodation.
For the quantitative detection of human IL-12/IL-23 p40.
Novus Biologicals™ Human IL12B ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Human IL-12 p70 Quantikine HS ELISA Kit, R&D Systems™
a 6.5 hour solid phase ELISA designed to measure IL-12 in cell culture supernates, serum, and plasma.
Invitrogen™ Mouse IL-12 (total) ELISA Kits Non-distribution product as customer accommodation.
Measure levels of total interleukin-12 (p40 + p70) in mouse serum, plasma and culture supernatant samples using 96-well plates.
R&D Systems Porcine IL-12/IL-23 p40 Quantikine ELISA Kit
a 4.5 hour solid-phase ELISA designed to measure porcine IL-12/IL-23 p40 in cell culture supernates, serum and plasma.
Novus Biologicals™ Human IL-12 ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Invitrogen™ Porcine IL-12 (IL12A) ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay porcine Porcine IL-12 (IL12A) levels in cell culture supernatants, plasma, serum samples
Mouse IL-12/IL-23 p40 Allele-specific Quantikine ELISA Kit, R&D Systems™
a 4.5 hour solid phase ELISA designed to measure mouse p40 subunit in IL-12 p70, IL-23, p40 homodimer, and p40 monomer in cell culture supernates and mouse serum.
Invitrogen™ eBioscience™ Mouse IL-12 Platinum ELISA Kit Non-distribution product as customer accommodation.
For the quantitative detection of mouse IL-12.
Novus Biologicals™ Primate IL-12 ELISA Kit [Biotin]
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Novus Biologicals™ Mouse IL-12 ELISA Kit [Biotin]
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Mouse IL-12/IL-23 p40 NonAllele-specific Quantikine ELISA Kit, R&D Systems™
a 4.5 hour solid phase ELISA designed to measure mouse p40 subunit in IL-12 p70, IL-23, p40 homodimer, and p40 monomer in cell culture supernates, mouse serum, and plasma.
eBioscience™ Monkey IL-12 (p70) Platinum ELISA Kit Non-distribution product as customer accommodation.
For the quantitative detection of monkey IL-12 p70.
Novus Biologicals™ Human IL-12 ELISA Kit [Biotin]
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
  spinner