IL-2 ELISA Kits
R&D Systems™ Human IL-2 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
A 4.5 hour solid phase ELISA designed to measure IL-2 levels in cell culture supernates, serum, and plasma. It contains E. coli-expressed recombinant human IL-2 and antibodies raised against recombinant human IL-2.
Invitrogen™ Human IL-2 ELISA Kits Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Interleukin 2 Recombinant protein SIL2 from Thermo Scientific™ specifically detects Interleukin 2 in samples, and it is validated for ELISA and Western Blot
R&D Systems™ Mouse IL-2 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid phase ELISA designed to measure mouse IL-2 levels in cell culture supernates and serum.
eBioscience™ Mouse IL-2 ELISA Ready-SET-Go!™ Kit Non-distribution product as customer accommodation.
Specifically engineered for accurate and precise measurement of mouse IL-2 protein levels from samples including serum, and cell culture supernatants.
R&D Systems™ Rabbit IL-2 DuoSet, 15 Plate
Sufficient for at least 15 96-well plates
eBioscience™ Human IL-2 ELISA Ready-SET-Go!™ Kit, 2nd Generation Non-distribution product as customer accommodation.
Specifically engineered for accurate and precise measurement of human IL-2 protein levels from samples including serum, plasma, and cell culture supernatants. This second generation kit has increased sensitivity with a range from 2 to 250pg/mL.
eBioscience™ Human IL-2 Platinum ELISA Kit Non-distribution product as customer accommodation.
For the quantitative detection of human IL-2.
eBioscience™ Mouse IL-2 Platinum ELISA Kit Non-distribution product as customer accommodation.
For the quantitative detection of mouse IL-2.
R&D Systems™ Human IL-2 QuantiGlo ELISA Kit, R&D Systems™
a 5.5 hour solid phase ELISA designed to measure IL-2 levels in cell culture supernates, serum, and plasma.
Novus Biologicals™ Primate IL-2 ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Invitrogen™ eBioscience™ Human IL-2 Instant ELISA Kit Non-distribution product as customer accommodation.
For the quantitative detection of human Interleukin-2.
R&D Systems™ Rat IL-2 Quantikine ELISA Kit, R&D Systems™
a 4.5 hour solid phase ELISA designed to measure rat IL-2 levels in cell culture supernates, serum, and plasma.
Novus Biologicals™ Rat IL-2 ELISA Kit [Biotin]
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
eBioscience™ Monkey IL-2 Platinum ELISA Kit Non-distribution product as customer accommodation.
For the quantitative detection of monkey IL-2.
Invitrogen™ Bovine IL-2 ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay bovine Bovine IL-2 levels in cell culture supernatants, plasma, serum samples
Invitrogen™ Equine IL-2 ELISA Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Assay equine IL-2 levels in cell culture supernatants, plasma, serum samples
Novus Biologicals™ IL-2 ELISA Kit [Biotin]
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Novus Biologicals™ IL-2 ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
eBioscience™ Rat IL-2 Platinum ELISA Kit Non-distribution product as customer accommodation.
For the quantitative detection of rat IL-2.
Interleukin 2 Recombinant Protein, Invitrogen™
Interleukin 2 Recombinant Protein, SIL2, from Invitrogen™
  spinner