In-Cell ELISA Kits for Specific Targets
R&D Systems™ Human IGF-I Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 3.5 hour solid-phase ELISA designed to measure human IGF-I in cell culture supernates, serum, and plasma.
R&D Systems™ Human IL-15 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.25 or 4.5 hour solid phase ELISA designed to measure human IL-15 in cell culture supernates, serum, and plasma.
R&D Systems™ Human CXCL10/IP-10 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid-phase ELISA designed to measure human IP-10 in cell culture supernates, serum, plasma, saliva, and urine.
R&D Systems™ Human RBP4 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 2.5 hour solid phase ELISA designed to measure human RBP4 in cell culture supernates, serum, plasma, urine, and saliva.
R&D Systems™ Human C-Reactive Protein/CRP Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid-phase ELISA designed to measure human CRP in cell culture supernates, serum, and plasma.
R&D Systems™ Human TGF-beta 1 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
A 4.5 hour solid phase ELISA designed to measure TGF-beta 1 in acid activated cell culture supernates, serum, plasma, and urine. It contains recombinant human TGF-beta 1 expressed by CHO cells
R&D Systems™ Human IL-7 Quantikine HS ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a solid phase ELISA that specifically measures IL-7 in 18-24 hours (with an overnight incubation step). This kit is designed to measure IL-7 levels in cell culture supernates, serum, and plasma.
R&D Systems™ Human sCD14 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
A 4.5-hour solid-phase ELISA designed to measure human soluble CD14 in cell culture supernants, serum and plasma.
eBioscience™ Mouse IFN gamma Platinum ELISA Kit, extra sensitive Non-distribution product as customer accommodation.
For the quantitative detection of mouse IFN-gamma.
R&D Systems™ Human Erythropoietin Quantikine IVD ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
Designed to measure Epo levels in serum or plasma in less than 4.5 hours (or less than 2.5 hours using the shaker protocol) for the determination of erythropoietin (Epo) concentration in human serum and plasma as an aid in the diagnosis of anemia and polycythemia. For in vitro diagnostic use.
R&D Systems™ Mouse Erythropoietin Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid-phase ELISA designed to measure mouse Epo in cell culture supernates, tissue homogenates, serum, and plasma.
R&D Systems™ Human Total Adiponectin/Acrp30 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid-phase ELISA designed to measure total (low, middle, and high molecular weight) human Adiponectin in cell culture supernates, serum, and plasma.
R&D Systems™ Mouse Lipocalin-2/NGAL Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid phase ELISA designed to measure mouse Lipocalin-2 in cell culture supernates, mouse serum, platelet-poor plasma, and urine.
Invitrogen™ IL-2 Mouse ELISA Kit, High Sensitivity Non-distribution product as customer accommodation.
High Sensitivity Sandwich ELISA Kit
R&D Systems™ Human B7-H3 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
Sufficient for at least one 96-well plates
R&D Systems™ Human/Mouse/Rat Activin A Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 5.0 hour solid-phase ELISA designed to measure Activin A in human, mouse, or rat cell culture supernates, serum, plasma, and saliva.
R&D Systems™ Human TIM-1/KIM-1/HAVCR Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid phase ELISA designed to measure TIM-1 in urine.
R&D Systems™ Prostaglandin E2 Parameter Assay Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 3.5 hour forward sequential competitive enzyme immunoassay designed to measure PGE2 in cell culture supernates, serum, plasma, and urine
Invitrogen™ Novex™ NP40 Cell Lysis Buffer Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Suitable for preparation of cell extracts
R&D Systems™ Human Leptin Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 3.5 hour solid phase ELISA designed to measure soluble human Leptin in cell culture supernates, serum, and plasma.
R&D Systems™ Human GDF-15 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 3.5 hour solid phase ELISA designed to measure GDF-15 in cell culture supernates, serum, plasma, and urine.
R&D Systems™ Creatinine Parameter Assay Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
Sufficient for at least two 96-well plates
R&D Systems™ Testosterone Parameter Assay Kit, R&D Systems™
a 4.5 hour competitive enzyme immunoassay designed to measure Testosterone in cell culture supernates, serum, and plasma
eBioscience™ Mouse IFN alpha Platinum ELISA Kit Non-distribution product as customer accommodation.
For the quantitative detection of mouse IFN-alpha.
Zeptometrix RETROtek HIV-1 p24 Antigen ELISA Available on GSA/VA Contract for Federal Government customers only.
Detects Human Immunodeficiency Virus Type 1 (HIV-1) p24 antigen in research specimens
R&D Systems™ Porcine CXCL8/IL-8 Quantikine ELISA Kit, R&D Systems™
a 4.5 hour solid phase ELISA designed to measure porcine IL-8 in cell culture supernates and serum.
R&D Systems™ Human FGF-21 Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.5 hour solid-phase ELISA designed to measure human FGF-21 in cell culture supernates, serum, and plasma.
R&D Systems™ Human Free BDNF Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 3.5 hour solid phase ELISA designed to measure human BDNF in cell culture supernates, serum, and plasma.
  spinner